Search
Search

Entrez Gene ID: | 81554 |
Gene Name: | RCC1 like |
Gene Aliases: |
WBSCR16 |
Location: |
Chr.7:75027119-75073802 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RCC1L | NM_001281441.2 | NP_001268370.1 | ||
| NM_001363447.2 | NP_001350376.1 | |||
| NM_030798.5 | NP_110425.2 | |||
| NM_148842.3 | NP_683682.1 | |||
| AF410455.1 | AAM62304.1 | |||
| AK309117.1 | ||||
| AL136804.1 | CAB66738.1 | |||
| AL536085.3 | ||||
| BC007823.2 | AAH07823.1 | |||
| BC019008.2 | AAH19008.1 | |||
| BC032712.1 | AAH32712.1 | |||
| BC040695.1 | AAH40695.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3219312 | Chr.7:75040022 - 75094199 on Build GRCh38 | Insertion |
|
| nsv1032137 | Chr.7:74697471 - 75115763 on Build GRCh38 | Gain |
|
| nsv4151648 | Chr.7:75058099 - 75075162 on Build GRCh38 | Duplication |
|
| nsv8145 | Chr.7:74961729 - 75076679 on Build GRCh38 | Gain |
|
| esv3613719 | Chr.7:75058352 - 75077912 on Build GRCh38 | Gain |
|
| nsv6607142 | Chr.7:75064633 - 75069155 on Build GRCh38 | Deletion |
|
| nsv5474250 | Chr.7:75058139 - 75077492 on Build GRCh38 | Duplication |
|
Set Membership: |
|