Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04379258_cn
          See other PRPF18 CNV Assays ›
          Gene Symbol
          PRPF18
          Assay Reference Genome
          Location

          Chr.10:13592491 on build GRCh38
          Cytoband
          Assay ID Hs04379258_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          8559

          Gene Name:

          pre-mRNA processing factor 18

          Gene Aliases:

          PRP18, hPrp18

          Location:

          Chr.10:13586965-13655929 on Build GRCh38

          Assay Gene Location:

          Within Intron 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PRPF18 NM_001395875.1 NP_001382804.1
          NM_001395876.1 NP_001382805.1
          NM_001395877.1 NP_001382806.1
          NM_001395878.1 NP_001382807.1
          NM_001395879.1 NP_001382808.1
          NM_001395880.1 NP_001382809.1
          NM_001395881.1 NP_001382810.1
          NM_001395882.1 NP_001382811.1
          NM_001395883.1 NP_001382812.1
          NM_001395884.1 NP_001382813.1
          NM_001395885.1 NP_001382814.1
          NM_001395886.1 NP_001382815.1
          NM_001395887.1 NP_001382816.1
          NM_003675.4 NP_003666.1
          NR_172904.1
          NR_172905.1
          NR_172906.1
          XM_047425923.1 XP_047281879.1
          XM_047425924.1 XP_047281880.1
          XM_047425925.1 XP_047281881.1
          XM_047425926.1 XP_047281882.1
          XM_047425927.1 XP_047281883.1
          XM_047425928.1 XP_047281884.1
          XM_047425929.1 XP_047281885.1
          XM_047425930.1 XP_047281886.1
          XM_047425931.1 XP_047281887.1
          XM_047425932.1 XP_047281888.1
          XM_047425933.1 XP_047281889.1
          XM_047425934.1 XP_047281890.1
          XM_047425935.1 XP_047281891.1
          XM_047425936.1 XP_047281892.1
          XM_047425937.1 XP_047281893.1
          XM_047425938.1 XP_047281894.1
          XM_047425939.1 XP_047281895.1
          XM_047425940.1 XP_047281896.1
          XM_047425941.1 XP_047281897.1
          XM_047425942.1 XP_047281898.1
          XM_047425943.1 XP_047281899.1
          XM_047425944.1 XP_047281900.1
          XM_047425945.1 XP_047281901.1
          XM_047425946.1 XP_047281902.1
          XM_047425947.1 XP_047281903.1
          XM_047425948.1 XP_047281904.1
          XM_047425949.1 XP_047281905.1
          XM_047425950.1 XP_047281906.1
          XM_047425951.1 XP_047281907.1
          XM_047425952.1 XP_047281908.1
          XM_047425953.1 XP_047281909.1
          XM_047425954.1 XP_047281910.1
          XM_047425955.1 XP_047281911.1
          XM_047425956.1 XP_047281912.1
          XM_047425957.1 XP_047281913.1
          XM_047425958.1 XP_047281914.1
          XM_047425959.1 XP_047281915.1
          XM_047425960.1 XP_047281916.1
          XM_047425961.1 XP_047281917.1
          XM_047425962.1 XP_047281918.1
          XM_047425963.1 XP_047281919.1
          XM_047425964.1 XP_047281920.1
          XM_047425965.1 XP_047281921.1
          XM_047425966.1 XP_047281922.1
          XM_047425967.1 XP_047281923.1
          AK314814.1 BAG37337.1
          AU142253.1
          BC000794.1 AAH00794.1
          U51990.1 AAB41490.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv947743 Chr.10:13591190 - 13592776 on Build GRCh38 Duplication PRPF18
          nsv4188258 Chr.10:13543727 - 13697859 on Build GRCh38 Duplication PRPF18 FRMD4A
          nsv4175718 Chr.10:13580085 - 13654763 on Build GRCh38 Duplication PRPF18 FRMD4A
          nsv831790 Chr.10:13536081 - 13716895 on Build GRCh38 Loss PRPF18 FRMD4A
          esv3891761 Chr.10:13544627 - 13695972 on Build GRCh38 Gain PRPF18 FRMD4A
          esv2761575 Chr.10:13581531 - 13659502 on Build GRCh38 Gain PRPF18 FRMD4A

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          generation of catalytic spliceosome for second transesterification step
          mRNA processing
          RNA splicing
          protein binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline