Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04502391_cn
          See other CYP2D6 CNV Assays ›
          Gene Symbol
          CYP2D6
          Assay Reference Genome
          Location

          Chr.22:42127680 on build GRCh38
          Cytoband
          Assay ID Hs04502391_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          1565

          Gene Name:

          cytochrome P450 family 2 subfamily D member 6 (gene/pseudogene)

          Gene Aliases:

          CPD6, CYP2D, CYP2D7AP, CYP2D7BP, CYP2D7P2, CYP2D8P2, CYP2DL1, CYPIID6, P450-DB1, P450C2D, P450DB1

          Location:

          Chr.22:48826-53137 on Build GRCh38

          Assay Gene Location:

          Within Intron 6
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          CYP2D6 NM_000106.6 NP_000097.3
          NM_001025161.3 NP_001020332.2
          AK309600.1 4 1272
          AY663390.1 AAU87043.1
          BC066877.1 AAH66877.1
          BC067432.1 AAH67432.1
          BC075023.2 AAH75023.1
          BC075024.2 AAH75024.1
          BC106757.1 AAI06758.1
          BC106758.1 AAI06759.1
          BC126858.1 AAI26859.1
          CK032845.1
          CR456430.1 CAG30316.1
          M20403.1 AAA52153.1
          X07618.1
          X08006.1 CAA30807.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv3642 Chr.22:42115497 - 42128296 on Build GRCh38 Insertion CYP2D6 NDUFA6-DT
          dgv1319e212 Chr.22:42126516 - 42145482 on Build GRCh38 Gain CYP2D6 CYP2D7
          esv6632 Chr.22:42125966 - 42139888 on Build GRCh38 Loss CYP2D6 CYP2D7
          nsv6048521 Chr.22:42125965 - 42135344 on Build GRCh38 Deletion CYP2D6
          esv3568417 Chr.22:42126951 - 42144107 on Build GRCh38 Loss CYP2D6 CYP2D7
          nsv834210 Chr.22:42057329 - 42209100 on Build GRCh38 Loss TCF20 NDUFA6 SMDT1 SNORD13P1 NAGA WBP2NL CYP2D6 CYP2D7 NDUFA6-DT PHETA2
          nsv829243 Chr.22:42121254 - 42154514 on Build GRCh38 Loss CYP2D6 CYP2D7 NDUFA6-DT
          esv33893 Chr.22:42126620 - 42142218 on Build GRCh38 Gain+Loss CYP2D6 CYP2D7
          nsv436350 Chr.22:42125831 - 42135447 on Build GRCh38 Deletion CYP2D6
          nsv5954144 Chr.22:42121553 - 42132728 on Build GRCh38 Deletion CYP2D6 NDUFA6-DT
          nsv3641 Chr.22:42099466 - 42146296 on Build GRCh38 Deletion CYP2D6 CYP2D7 NDUFA6-DT
          dgv4710n223 Chr.22:42125201 - 42135000 on Build GRCh38 Duplication CYP2D6 NDUFA6-DT
          nsv955164 Chr.22:42122897 - 42144299 on Build GRCh38 Deletion CYP2D6 CYP2D7 NDUFA6-DT
          nsv5585870 Chr.22:42125912 - 42138063 on Build GRCh38 Deletion CYP2D6
          nsv498989 Chr.22:42123210 - 42135371 on Build GRCh38 Loss CYP2D6 NDUFA6-DT
          dgv4711n223 Chr.22:42127556 - 42142900 on Build GRCh38 Deletion CYP2D6 CYP2D7
          esv2171396 Chr.22:42125911 - 42139869 on Build GRCh38 Deletion CYP2D6 CYP2D7
          nsv6541771 Chr.22:42125701 - 42129900 on Build GRCh38 Duplication CYP2D6
          esv3575484 Chr.22:42118886 - 42144107 on Build GRCh38 Gain CYP2D6 CYP2D7 NDUFA6-DT
          nsv508737 Chr.22:42052007 - 42134336 on Build GRCh38 Insertion NDUFA6 SMDT1 SNORD13P1 NAGA WBP2NL CYP2D6 NDUFA6-DT PHETA2
          nsv6552980 Chr.22:42127201 - 42129800 on Build GRCh38 Duplication CYP2D6
          nsv1110402 Chr.22:42125795 - 42137591 on Build GRCh38 Duplication CYP2D6
          nsv6535893 Chr.22:41572303 - 44198988 on Build GRCh38 Duplication SCUBE1-AS1 EFCAB6 PMM1 SNORD13P1 XRCC6 POLDIP3 TTLL1-AS1 SCUBE1 MEI1 CCDC134 WBP2NL CYP2D6 CENPM SULT4A1 PNPLA3 MIR378I NDUFA6-DT C22orf46P SNU13 TCF20 EFCAB6-DT LOC107985567 SAMM50 NFAM1 CYP2D7 SERHL2 SREBF2 LINC01639 ATP5MGL SERHL PHETA2 SEPTIN3 EFCAB6-AS1 NAGA TNFRSF13C RRP7A SMIM45 TTLL1 NDUFA6 SMDT1 DESI1 SHISA8 A4GALT PARVG ARFGAP3 CSDC2 TTLL12 MIR33A BIK MCAT LINC01315 SREBF2-AS1 MPPED1 PACSIN2 CYB5R3 RRP7BP PNPLA5 OGFRP1 LOC101927344 TSPO SCUBE1-AS2 RNU12 PARVB
          nsv6052872 Chr.22:42125962 - 42139826 on Build GRCh38 Duplication CYP2D6 CYP2D7
          esv2724275 Chr.22:42123193 - 42141302 on Build GRCh38 Deletion CYP2D6 CYP2D7 NDUFA6-DT
          nsv3231826 Chr.22:42118380 - 42147472 on Build GRCh38 Insertion CYP2D6 CYP2D7 NDUFA6-DT

          Back To Top

          More Information


          Important Information

          Individuals may carry null alleles or extra copies of CYP2D6. Some CYP2D6 alleles contain sequences derived from the highly homologous CYP2D7 pseudogene. Hs04502391_cn specifically targets CYP2D6 intron 6 sequences and will not amplify CYP2D7 or CYP2D8 pseudogenes, but will amplify CYP2D6/CYP2D7 hybrid alleles carrying CYP2D6 intron 6 sequences (e.g. CYP2D6*36 contains conversion to CYP2D7 in exon 9).

          Additional Information:

          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation Validated

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          fatty acid metabolic process
          xenobiotic metabolic process
          steroid metabolic process
          cholesterol metabolic process
          estrogen metabolic process
          coumarin metabolic process
          alkaloid metabolic process
          alkaloid catabolic process
          monoterpenoid metabolic process
          arachidonic acid metabolic process
          isoquinoline alkaloid metabolic process
          xenobiotic catabolic process
          retinol metabolic process
          long-chain fatty acid biosynthetic process
          oxidative demethylation
          negative regulation of cellular organofluorine metabolic process
          monooxygenase activity
          iron ion binding
          oxidoreductase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
          heme binding
          metal ion binding
          anandamide 8,9 epoxidase activity
          anandamide 11,12 epoxidase activity
          anandamide 14,15 epoxidase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline