Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04506824_cn
          See other LARGE1 CNV Assays ›
          Gene Symbol
          LARGE1
          Assay Reference Genome
          Location

          Chr.22:33795372 on build GRCh38
          Cytoband
          Assay ID Hs04506824_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          9215

          Gene Name:

          LARGE xylosyl- and glucuronyltransferase 1

          Gene Aliases:

          LARGE, MDC1D, MDDGA6, MDDGB6

          Location:

          Chr.22:33066663-33922824 on Build GRCh38

          Assay Gene Location:

          Within Intron 7
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          LARGE1 NM_001362949.2 NP_001349878.1
          NM_001362951.2 NP_001349880.1
          NM_001362953.2 NP_001349882.1
          NM_001378624.1 NP_001365553.1
          NM_001378625.1 NP_001365554.1
          NM_001378626.1 NP_001365555.1
          NM_001378627.1 NP_001365556.1
          NM_001378628.1 NP_001365557.1
          NM_001378629.1 NP_001365558.1
          NM_004737.7 NP_004728.1
          NM_133642.5 NP_598397.1
          XM_024452302.2 XP_024308070.1
          XM_024452303.2 XP_024308071.1
          XM_047441599.1 XP_047297555.1
          XM_047441600.1 XP_047297556.1
          XM_047441601.1 XP_047297557.1
          XM_047441602.1 XP_047297558.1
          XM_047441604.1 XP_047297560.1
          XM_047441605.1 XP_047297561.1
          AB011181.3 BAA25535.3
          AI199197.1
          AJ007583.1 CAA07571.1
          AK294766.1 BAH11875.1
          AK295636.1 BAH12135.1
          BF057493.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv2758833 Chr.22:33773808 - 33807492 on Build GRCh38 Loss LARGE1
          dgv4691n223 Chr.22:33699218 - 33802914 on Build GRCh38 Deletion LARGE1 SNORA50B LARGE-AS1
          esv3647634 Chr.22:33795330 - 33797659 on Build GRCh38 Loss LARGE1
          nsv5540792 Chr.22:33795381 - 33797706 on Build GRCh38 Deletion LARGE1
          nsv459878 Chr.22:33710759 - 33830604 on Build GRCh38 Loss LARGE1 LARGE-AS1
          nsv6548162 Chr.22:33787559 - 33825116 on Build GRCh38 Deletion LARGE1
          nsv6042341 Chr.22:33735842 - 33858438 on Build GRCh38 Deletion LARGE1 LARGE-AS1
          esv2763261 Chr.22:33435362 - 33892401 on Build GRCh38 Loss LARGE1 SNORA50B LARGE-AS1 MIR4764
          esv3568379 Chr.22:33458829 - 33815605 on Build GRCh38 Loss LARGE1 SNORA50B LARGE-AS1
          nsv1055203 Chr.22:33702431 - 33800698 on Build GRCh38 Loss LARGE1 SNORA50B LARGE-AS1
          nsv524156 Chr.22:33749609 - 33864750 on Build GRCh38 Loss LARGE1 LARGE-AS1

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel development
          behavioral fear response
          neuron migration
          N-acetylglucosamine metabolic process
          protein glycosylation
          protein O-linked glycosylation
          protein targeting to membrane
          glycosphingolipid biosynthetic process
          water transport
          intracellular protein transport
          striated muscle contraction
          plasma membrane organization
          cytoskeleton organization
          chemical synaptic transmission
          neuromuscular synaptic transmission
          brain development
          heart development
          muscle organ development
          neuromuscular junction development
          sensory perception of sound
          memory
          protein localization
          determination of adult lifespan
          glycoprotein metabolic process
          glycoprotein biosynthetic process
          response to light stimulus
          response to mechanical stimulus
          gene expression
          retina layer formation
          myotube differentiation
          dentate gyrus development
          nerve development
          principal sensory nucleus of trigeminal nerve development
          macrophage differentiation
          post-embryonic hindlimb morphogenesis
          multicellular organism growth
          protein O-linked mannosylation
          protein modification process
          myelination
          skeletal muscle tissue regeneration
          post-translational protein modification
          muscle cell cellular homeostasis
          regulation of synaptic plasticity
          astrocyte differentiation
          cardiac muscle tissue development
          skeletal muscle fiber development
          synapse organization
          neuromuscular process controlling posture
          neuromuscular process
          synaptic growth at neuromuscular junction
          localization of cell
          striated muscle cell development
          cardiac muscle cell development
          transmembrane transport
          retina development in camera-type eye
          limb development
          long-term synaptic potentiation
          bone development
          skeletal muscle organ development
          retina vasculature development in camera-type eye
          connective tissue development
          protein-containing complex assembly
          basement membrane organization
          potassium ion transmembrane transport
          protein localization to membrane
          protein localization to plasma membrane
          walking behavior
          acetylcholine receptor signaling pathway
          skeletal muscle fiber differentiation
          reactive gliosis
          catalytic activity
          protein binding
          acetylglucosaminyltransferase activity
          glucuronosyltransferase activity
          transferase activity
          transferase activity, transferring glycosyl groups
          transferase activity, transferring hexosyl groups
          manganese ion binding
          UDP-xylosyltransferase activity
          xylosyltransferase activity
          metal ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline