Search
Search

Entrez Gene ID: | 1852 |
Gene Name: | dual specificity phosphatase 9 |
Gene Aliases: |
MKP-4, MKP4 |
Location: |
Chr.X:153642441-153651326 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DUSP9 | NM_001395.4 | NP_001386.1 | ||
| XM_011531123.2 | 1 | 846 | XP_011529425.1 | |
| AK314250.1 | BAG36916.1 | |||
| BC034936.1 | AAH34936.1 | |||
| BC042166.1 | AAH42166.1 | |||
| MN308287.1 | ||||
| Y08302.1 | CAA69610.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2666278 | Chr.X:153641591 - 153644638 on Build GRCh38 | Deletion |
|
| esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss |
|
| nsv4038621 | Chr.X:153640546 - 153673645 on Build GRCh38 | Deletion |
|