Search
Search

Entrez Gene ID: | 3020 |
Gene Name: | H3 histone, family 3A |
Gene Aliases: |
H3.3A, H3F3 |
Location: |
Chr.1:226062707-226072002 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| H3F3A | NM_002107.4 | NP_002098.1 | ||
| AK311905.1 | ||||
| BC029405.1 | AAH29405.1 | |||
| BC095447.1 | AAH95447.1 | |||
| BM760856.1 | ||||
| M11353.1 | AAA52654.1 | |||
| M11354.1 | AAA52653.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3589008 | Chr.1:226070529 - 226075723 on Build GRCh38 | Gain |
|
| nsv832747 | Chr.1:226006442 - 226185714 on Build GRCh38 | Gain |
|
| nsv952128 | Chr.1:226064300 - 226073399 on Build GRCh38 | Duplication |
|
| nsv945330 | Chr.1:226071371 - 226072183 on Build GRCh38 | Duplication |
|
Set Membership: |
|