Search
Search

Entrez Gene ID: | 5581 |
Gene Name: | protein kinase C epsilon |
Gene Aliases: |
PKCE, nPKC-epsilon |
Location: |
Chr.2:45651315-46187990 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKCE | NM_005400.2 | NP_005391.1 | ||
| XM_005264428.1 | XP_005264485.1 | |||
| XM_005264431.3 | XP_005264488.1 | |||
| XM_011532971.2 | XP_011531273.1 | |||
| XM_011532975.2 | XP_011531277.1 | |||
| XM_011532978.1 | XP_011531280.1 | |||
| XM_011532980.2 | XP_011531282.1 | |||
| XM_017004486.1 | XP_016859975.1 | |||
| XM_017004487.1 | XP_016859976.1 | |||
| XM_017004488.1 | XP_016859977.1 | |||
| XM_017004489.1 | XP_016859978.1 | |||
| XM_017004490.1 | XP_016859979.1 | |||
| XM_017004492.1 | XP_016859981.1 | |||
| AK313842.1 | ||||
| AL832345.1 | ||||
| BC109033.2 | AAI09034.1 | |||
| BC109034.2 | AAI09035.1 | |||
| X65293.1 | CAA46388.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3590595 | Chr.2:45796324 - 45804310 on Build GRCh38 | Loss |
|
| esv2668866 | Chr.2:45796257 - 45804171 on Build GRCh38 | Deletion |
|
| esv2720011 | Chr.2:45792872 - 45810252 on Build GRCh38 | Deletion |
|
| esv2762545 | Chr.2:45419685 - 46137453 on Build GRCh38 | Gain |
|
| nsv581742 | Chr.2:45794467 - 45825358 on Build GRCh38 | Loss |
|
| nsv2713 | Chr.2:45776481 - 45805904 on Build GRCh38 | Deletion |
|
Set Membership: |
|