Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04611393_cn
          See other RAMP1 CNV Assays ›
          Gene Symbol
          RAMP1
          Assay Reference Genome
          Location

          Chr.2:237909216 on build GRCh38
          Cytoband
          Assay ID Hs04611393_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          10267

          Gene Name:

          receptor activity modifying protein 1

          Gene Aliases:

          -

          Location:

          Chr.2:237858880-237912106 on Build GRCh38

          Assay Gene Location:

          Within Intron 5
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          RAMP1 NM_001308353.2 NP_001295282.1
          NM_005855.4 NP_005846.1
          XM_017003152.3 XP_016858641.1
          XM_017003153.3 XP_016858642.1
          XM_017003156.3 XP_016858645.1
          AI017108.1
          AJ001014.1 CAA04472.1
          AY265457.1 AAP23298.1
          BC000548.2 AAH00548.1
          BI548986.1
          BM978161.1
          CR542032.1 CAG46829.1
          CR542044.1 CAG46841.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv4075099 Chr.2:237888275 - 237918497 on Build GRCh38 Duplication RAMP1
          nsv508205 Chr.2:237889539 - 237970025 on Build GRCh38 Deletion UBE2F UBE2F-SCLY RAMP1
          nsv1000252 Chr.2:235899999 - 238766417 on Build GRCh38 Loss ACKR3 ILKAP LRRFIP1 AGAP1 ERFE SCLY MLPH TRAF3IP1 GBX2 ESPNL ASB1 COPS8 TARDBPP3 DRC11 IQCA1-AS1 RBM44 ASB18 COPS8-DT UBE2F-SCLY HES6 LOC93463 RAB17-DT KLHL30 RAB17 LINC01107 LINC02610 PER2 PRLH GBX2-AS1 UBE2F MIR6811 RAMP1 COL6A3
          nsv6337133 Chr.2:237890456 - 237925443 on Build GRCh38 Duplication RAMP1

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          calcium ion transport
          intracellular protein transport
          G-protein coupled receptor signaling pathway
          adenylate cyclase-activating G-protein coupled receptor signaling pathway
          regulation of G-protein coupled receptor protein signaling pathway
          protein transport
          receptor internalization
          cellular response to hormone stimulus
          positive regulation of protein glycosylation
          protein localization to plasma membrane
          amylin receptor signaling pathway
          amylin receptor 1 signaling pathway
          calcitonin gene-related peptide receptor signaling pathway
          calcitonin gene-related peptide receptor activity
          protein binding
          coreceptor activity
          amylin receptor activity
          calcitonin gene-related peptide binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline