Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04711067_cn
          See other STK39 CNV Assays ›
          Gene Symbol
          STK39
          Assay Reference Genome
          Location

          Chr.2:168202816 on build GRCh38
          Cytoband
          Assay ID Hs04711067_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          27347

          Gene Name:

          serine/threonine kinase 39

          Gene Aliases:

          DCHT, PASK, SPAK

          Location:

          Chr.2:167954022-168247595 on Build GRCh38

          Assay Gene Location:

          Within Intron 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          STK39 NM_001410961.1 NP_001397890.1
          NM_013233.3 NP_037365.2
          XM_017003813.3 XP_016859302.1
          XM_017003814.3 XP_016859303.1
          XM_017003816.3 XP_016859305.1
          XM_047443941.1 XP_047299897.1
          XM_047443944.1 XP_047299900.1
          AF030403.1 AAD01901.1
          AF099989.1 AAC72238.1
          KJ534948.1 AHW56588.1
          KJ534949.1 AHW56589.1
          KJ534950.1 AHW56590.1
          KJ534951.1 AHW56591.1
          KJ534999.1 AHW56639.1
          KJ535059.1 AHW56698.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv3022 Chr.2:168201820 - 168233217 on Build GRCh38 Insertion STK39

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          response to dietary excess
          protein phosphorylation
          cell volume homeostasis
          inflammatory response
          signal transduction
          regulation of blood pressure
          positive regulation of T cell chemotaxis
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          cellular chloride ion homeostasis
          negative regulation of transmembrane transport
          intracellular signal transduction
          sodium ion transmembrane transport
          cellular response to potassium ion
          maintenance of lens transparency
          chemokine (C-X-C motif) ligand 12 signaling pathway
          macrophage activation
          positive regulation of potassium ion transport
          protein autophosphorylation
          regulation of inflammatory response
          ion homeostasis
          renal sodium ion absorption
          cellular hyperosmotic response
          cellular hypotonic response
          potassium ion transmembrane transport
          negative regulation of pancreatic juice secretion
          positive regulation of p38MAPK cascade
          negative regulation of potassium ion transmembrane transport
          negative regulation of sodium ion transmembrane transport
          positive regulation of potassium ion import
          positive regulation of sodium ion import across plasma membrane
          response to aldosterone
          regulation of cation transmembrane transport
          cellular response to chemokine
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          potassium channel inhibitor activity
          protein kinase binding
          ion channel binding
          ion channel regulator activity
          protein serine kinase activity
          transporter activator activity
          transporter inhibitor activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline