Search
Search

Entrez Gene ID: | 7325 |
Gene Name: | ubiquitin conjugating enzyme E2 E2 |
Gene Aliases: |
UBCH8 |
Location: |
Chr.3:23203098-23591925 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2E2 | NM_001370225.1 | NP_001357154.1 | ||
| NM_001370226.1 | NP_001357155.1 | |||
| NM_152653.4 | NP_689866.1 | |||
| XM_047448843.1 | XP_047304799.1 | |||
| XM_047448845.1 | XP_047304801.1 | |||
| AK057886.1 | BAB71605.1 | |||
| AK307075.1 | ||||
| BC022332.1 | AAH22332.1 | |||
| BI603911.1 | ||||
| DB476522.1 | ||||
| HY006728.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1007373 | Chr.3:23326461 - 23668840 on Build GRCh38 | Gain |
|
| nsv6373205 | Chr.3:23327004 - 23676760 on Build GRCh38 | Duplication |
|
| nsv6361959 | Chr.3:23193347 - 23376385 on Build GRCh38 | Duplication |
|
| nsv1002823 | Chr.3:23236347 - 23581592 on Build GRCh38 | Loss |
|
| dgv4702n100 | Chr.3:23366936 - 23427572 on Build GRCh38 | Loss |
|
| nsv1005186 | Chr.3:21989661 - 24089392 on Build GRCh38 | Gain |
|
Set Membership: |
|