Search
Search

Entrez Gene ID: | 23225 |
Gene Name: | nucleoporin 210 |
Gene Aliases: |
GP210, POM210 |
Location: |
Chr.3:13316230-13420319 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUP210 | NM_024923.3 | NP_079199.2 | ||
| XM_005264994.3 | XP_005265051.1 | |||
| XM_011533538.2 | XP_011531840.1 | |||
| XM_017006021.1 | XP_016861510.1 | |||
| AK026042.1 | BAB15332.1 | |||
| AK225409.1 | ||||
| BC067089.1 | AAH67089.1 |
Set Membership: |
|