Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04902228_cn
          See other PHACTR1 CNV Assays ›
          Gene Symbol
          PHACTR1
          Assay Reference Genome
          Location

          Chr.6:12862265 on build GRCh38
          Cytoband
          Assay ID Hs04902228_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          221692

          Gene Name:

          phosphatase and actin regulator 1

          Gene Aliases:

          DEE70, EIEE70, RPEL, RPEL1, dJ257A7.2

          Location:

          Chr.6:12716767-13287837 on Build GRCh38

          Assay Gene Location:

          Within Intron 5
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PHACTR1 NM_001242648.4 NP_001229577.1
          NM_001322308.3 NP_001309237.1
          NM_001322309.3 NP_001309238.1
          NM_001322310.2 NP_001309239.1
          NM_001374581.2 NP_001361510.1
          NM_001374582.1 NP_001361511.1
          NM_001374584.1 NP_001361513.1
          NM_030948.6 NP_112210.1
          XM_005248934.4 XP_005248991.1
          XM_017010459.3 XP_016865948.1
          XM_047418372.1 XP_047274328.1
          XM_047418373.1 XP_047274329.1
          XM_047418374.1 XP_047274330.1
          AB051520.1 BAB21824.1
          AK090769.1 BAG52223.1
          AK290017.1 BAF82706.1
          AK295263.1 BAG58252.1
          BC101526.1 AAI01527.1
          BC101528.1 AAI01529.1
          CR746945.1
          DA330657.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv1027534 Chr.6:12857816 - 12877365 on Build GRCh38 Loss PHACTR1
          nsv6411508 Chr.6:12861994 - 12862573 on Build GRCh38 Deletion PHACTR1
          dgv84n68 Chr.6:12769429 - 12972261 on Build GRCh38 Loss PHACTR1
          esv3890791 Chr.6:12790188 - 13033344 on Build GRCh38 Loss PHACTR1

          Back To Top

          More Information


          Additional Information:

          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation Validated

          Gene Ontology Categories:

          Function(s) Process(es)

          cerebral cortex development
          actin cytoskeleton organization
          actomyosin structure organization
          stress fiber assembly
          cell motility
          dendrite arborization
          regulation of neuron migration
          actin binding
          protein phosphatase inhibitor activity
          protein binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline