Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04925670_cn
          See other SUGCT CNV Assays ›
          Gene Symbol
          SUGCT
          Assay Reference Genome
          Location

          Chr.7:40310836 on build GRCh38
          Cytoband
          Assay ID Hs04925670_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          79783

          Gene Name:

          succinyl-CoA:glutarate-CoA transferase

          Gene Aliases:

          C7orf10, DERP13, GA3, ORF19

          Location:

          Chr.7:40135005-41038816 on Build GRCh38

          Assay Gene Location:

          Within Intron 10
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          SUGCT NM_001193311.2 NP_001180240.2
          NM_001193312.2 NP_001180241.2
          NM_001193313.2 NP_001180242.2
          NM_024728.3 NP_079004.2
          XM_006715775.4 XP_006715838.2
          XM_011515525.4 XP_011513827.2
          XM_011515526.2 XP_011513828.1
          XM_011515527.4 XP_011513829.2
          XM_011515529.4 XP_011513831.2
          XM_011515530.4 XP_011513832.2
          XM_017012621.1 XP_016868110.1
          XM_017012622.3 XP_016868111.2
          XM_047420840.1 XP_047276796.1
          XM_047420841.1 XP_047276797.1
          XM_047420842.1 XP_047276798.1
          XM_047420843.1 XP_047276799.1
          AB014767.1 BAB87807.1
          AK021870.1 BAB13922.1
          AK296048.1 BAG58812.1
          AK299133.1 BAG61185.1
          AK307935.1
          AK308764.1
          BC098117.1 AAH98117.1
          BC098261.1 AAH98261.1
          BC098310.1 AAH98310.1
          BC098318.1 AAH98318.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv1032278 Chr.7:40057772 - 40356701 on Build GRCh38 Gain MPLKIP CDK13 SUGCT
          nsv4133003 Chr.7:40181736 - 40362377 on Build GRCh38 Deletion SUGCT
          esv3891101 Chr.7:40263685 - 40320658 on Build GRCh38 Loss SUGCT
          nsv520748 Chr.7:40292919 - 40385541 on Build GRCh38 Loss SUGCT
          dgv11290n54 Chr.7:40153936 - 40320658 on Build GRCh38 Gain SUGCT
          nsv6609716 Chr.7:40251762 - 40319261 on Build GRCh38 Deletion SUGCT
          nsv4140988 Chr.7:40262712 - 40501511 on Build GRCh38 Deletion SUGCT
          nsv4136817 Chr.7:40290401 - 40390901 on Build GRCh38 Deletion SUGCT
          nsv4149820 Chr.7:40309047 - 40314253 on Build GRCh38 Duplication SUGCT
          nsv1017909 Chr.7:40297510 - 40388035 on Build GRCh38 Loss SUGCT
          nsv6606084 Chr.7:40290429 - 40391124 on Build GRCh38 Deletion SUGCT

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Process(es)

          catalytic activity
          transferase activity
          succinate-hydroxymethylglutarate CoA-transferase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline