Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04930522_cn
          See other SUGCT CNV Assays ›
          Gene Symbol
          SUGCT
          Assay Reference Genome
          Location

          Chr.7:40496633 on build GRCh38
          Cytoband
          Assay ID Hs04930522_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          79783

          Gene Name:

          succinyl-CoA:glutarate-CoA transferase

          Gene Aliases:

          C7orf10, DERP13, GA3, ORF19

          Location:

          Chr.7:40135005-41038816 on Build GRCh38

          Assay Gene Location:

          Within Intron 15
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          SUGCT NM_001193311.2 NP_001180240.2
          NM_001193312.2 NP_001180241.2
          NM_001193313.2 NP_001180242.2
          NM_024728.3 NP_079004.2
          XM_006715775.4 XP_006715838.2
          XM_011515525.4 XP_011513827.2
          XM_011515526.2 XP_011513828.1
          XM_011515527.4 XP_011513829.2
          XM_011515529.4 XP_011513831.2
          XM_017012621.1 XP_016868110.1
          XM_017012622.3 XP_016868111.2
          XM_047420840.1 XP_047276796.1
          XM_047420841.1 XP_047276797.1
          XM_047420842.1 XP_047276798.1
          XM_047420843.1 XP_047276799.1
          AB014767.1 BAB87807.1
          AK021870.1 BAB13922.1
          AK296048.1 BAG58812.1
          AK299133.1 BAG61185.1
          AK307935.1
          AK308764.1
          BC098117.1 AAH98117.1
          BC098261.1 AAH98261.1
          BC098310.1 AAH98310.1
          BC098318.1 AAH98318.1
          BM668297.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv6617288 Chr.7:40447731 - 40504946 on Build GRCh38 Deletion SUGCT
          nsv6601799 Chr.7:40400687 - 40584267 on Build GRCh38 Deletion SUGCT-AS1 SUGCT
          nsv6604807 Chr.7:40419091 - 40511015 on Build GRCh38 Deletion SUGCT
          nsv4140988 Chr.7:40262712 - 40501511 on Build GRCh38 Deletion SUGCT
          esv3612933 Chr.7:40496509 - 40498824 on Build GRCh38 Loss SUGCT
          nsv830969 Chr.7:40382977 - 40542790 on Build GRCh38 Gain SUGCT-AS1 SUGCT
          nsv5487536 Chr.7:40496528 - 40498816 on Build GRCh38 Deletion SUGCT
          nsv1029342 Chr.7:40449484 - 40503334 on Build GRCh38 Loss SUGCT
          nsv830971 Chr.7:40483476 - 40660573 on Build GRCh38 Gain+Loss SUGCT-AS1 SUGCT

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Process(es)

          catalytic activity
          transferase activity
          succinate-hydroxymethylglutarate CoA-transferase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline