Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04947094_cn
          See other FAM188B CNV Assays ›
          Gene Symbol
          FAM188B
          Assay Reference Genome
          Location

          Chr.7:30875081 on build GRCh38
          Cytoband
          7p14.3
          Assay ID Hs04947094_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          84182

          Gene Name:

          family with sequence similarity 188 member B

          Gene Aliases:

          C7orf67

          Location:

          Chr.7:30771417-30892387 on Build GRCh38

          Assay Gene Location:

          Within Intron 14
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          FAM188B NM_032222.2 NP_115598.2
          AF538962.1 AAQ10898.1
          AK090826.1
          BC098569.1 AAH98569.1

          Target Gene Details

          Entrez Gene ID:

          100526825

          Gene Name:

          INMT-FAM188B readthrough (NMD candidate)

          Gene Aliases:

          -

          Location:

          Chr.7:30752135-30892387 on Build GRCh38

          Assay Gene Location:

          Within Intron 16
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          INMT-FAM188B NR_037598.1
          AK308762.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv3571725 Chr.7:30874793 - 30876452 on Build GRCh38 Loss FAM188B INMT-FAM188B
          dgv6316n100 Chr.7:30609596 - 31145482 on Build GRCh38 Gain FAM188B GARS INMT-FAM188B ADCYAP1R1 AQP1 GHRHR CRHR2 INMT
          esv2761316 Chr.7:30615466 - 31127062 on Build GRCh38 Gain FAM188B GARS INMT-FAM188B ADCYAP1R1 AQP1 GHRHR CRHR2 INMT
          nsv527038 Chr.7:30617851 - 31122331 on Build GRCh38 Gain FAM188B GARS INMT-FAM188B ADCYAP1R1 AQP1 GHRHR CRHR2 INMT
          nsv436557 Chr.7:30288062 - 32730902 on Build GRCh38 Deletion INMT-FAM188B ZNRF2 ADCYAP1R1 DPY19L1P1 CRHR2 INMT LINC01176 FAM188B DKFZP586I1420 MIR550A1 GGCT AVL9 LOC401320 CCDC129 LSM5 NEUROD6 MIR550B1 NOD1 LOC101928268 PPP1R17 GARS LOC100130673 AQP1 GHRHR ZNRF2P1 PDE1C

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline