Search
Search

Entrez Gene ID: | 2770 |
Gene Name: | G protein subunit alpha i1 |
Gene Aliases: |
Gi |
Location: |
Chr.7:80134824-80219409 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNAI1 | NM_001256414.1 | NP_001243343.1 | ||
| NM_002069.5 | NP_002060.4 | |||
| AF055013.1 | AAC09361.1 | |||
| AF493905.1 | AAM12619.1 | |||
| AI878901.1 | ||||
| AK292953.1 | ||||
| AK304442.1 | ||||
| AL049933.1 | CAB43212.2 | |||
| BC026326.1 | AAH26326.1 | |||
| BT019775.1 | AAV38580.1 | |||
| M17219.1 | AAA52581.1 |
Set Membership: |
|