Search
Search

Entrez Gene ID: | 11112 |
Gene Name: | 3-hydroxyisobutyrate dehydrogenase |
Gene Aliases: |
NS5ATP1 |
Location: |
Chr.7:27525442-27662883 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIBADH | NM_001430749.1 | NP_001417678.1 | ||
| NM_152740.4 | NP_689953.1 | |||
| XM_047419834.1 | XP_047275790.1 | |||
| XM_047419835.1 | XP_047275791.1 | |||
| AB050000.1 | BAF42045.1 | |||
| AF529362.1 | AAQ09596.1 | |||
| AK316605.1 | BAG38192.1 | |||
| BC032324.1 | AAH32324.1 | |||
| GQ900978.1 | ADO22495.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830930 | Chr.7:27356219 - 27547736 on Build GRCh38 | Loss |
|
| nsv6607087 | Chr.7:27538514 - 27539208 on Build GRCh38 | Deletion |
|
Set Membership: |
|