Search
Search

Entrez Gene ID: | 8029 |
Gene Name: | cubilin |
Gene Aliases: |
IFCR, IGS, IGS1, MGA1, gp280 |
Location: |
Chr.10:16823966-17129811 on Build GRCh38 |
Assay Gene Location: | Within Intron 62 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CUBN | NM_001081.4 | NP_001072.2 | ||
| XM_011519709.3 | XP_011518011.1 | |||
| XM_011519710.3 | XP_011518012.1 | |||
| XM_011519711.4 | XP_011518013.1 | |||
| AB209880.1 | BAD93117.1 | |||
| AF034611.1 | AAC82612.1 | |||
| AK075228.1 | BAG52089.1 | |||
| AK308290.1 | ||||
| AK310404.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470924 | Chr.10:16864509 - 16951578 on Build GRCh38 | Gain |
|
| esv3891765 | Chr.10:16809409 - 16977014 on Build GRCh38 | Gain |
|
| nsv466758 | Chr.10:16672467 - 16954685 on Build GRCh38 | Gain |
|
| nsv831800 | Chr.10:16765669 - 16949419 on Build GRCh38 | Gain |
|
| nsv5481509 | Chr.10:16808100 - 16978218 on Build GRCh38 | Duplication |
|
| nsv1046088 | Chr.10:16786830 - 16906310 on Build GRCh38 | Gain |
|
| nsv5923254 | Chr.10:16671260 - 16954786 on Build GRCh38 | Duplication |
|
| esv3622477 | Chr.10:16808207 - 16934688 on Build GRCh38 | Gain |
|
| nsv5909086 | Chr.10:16761962 - 17211984 on Build GRCh38 | Deletion |
|
| nsv6435641 | Chr.10:16785481 - 16908110 on Build GRCh38 | Duplication |
|
Set Membership: |
|