Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05127426_cn
          See other ACSL5 CNV Assays ›
          Gene Symbol
          ACSL5
          Assay Reference Genome
          Location

          Chr.10:112391611 on build GRCh38
          Cytoband
          Assay ID Hs05127426_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          51703

          Gene Name:

          acyl-CoA synthetase long chain family member 5

          Gene Aliases:

          ACS2, ACS5, DIAR13, FACL5

          Location:

          Chr.10:112374116-112428376 on Build GRCh38

          Assay Gene Location:

          Within Intron 3
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          ACSL5 NM_001387037.1 NP_001373966.1
          NM_016234.4 NP_057318.2
          NM_203379.2 NP_976313.1
          NM_203380.2 NP_976314.1
          AB033899.1 BAA85979.1
          AK000339.1
          AK024932.1
          AK222782.1
          AK223288.1
          AL708480.1
          AY358520.1 AAQ88884.1
          BC007985.2 AAH07985.2
          CD689921.1
          CF595490.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv6446012 Chr.10:111938190 - 112596067 on Build GRCh38 Duplication TECTB MIR6715A VTI1A GPAM ZDHHC6 ACSL5 GUCY2GP MIR6715B
          nsv831990 Chr.10:112349267 - 112531997 on Build GRCh38 Loss VTI1A ZDHHC6 ACSL5 GUCY2GP

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          long-chain fatty acid metabolic process
          lipid metabolic process
          fatty acid metabolic process
          positive regulation of plasma membrane long-chain fatty acid transport
          long-chain fatty-acyl-CoA biosynthetic process
          regulation of extrinsic apoptotic signaling pathway
          nucleotide binding
          long-chain fatty acid-CoA ligase activity
          protein binding
          ATP binding
          ligase activity
          arachidonate-CoA ligase activity
          oleoyl-CoA ligase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline