Search
Search

Entrez Gene ID: | 221061 |
Gene Name: | family with sequence similarity 171 member A1 |
Gene Aliases: |
APCN, C10orf38 |
Location: |
Chr.10:15211643-15374554 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM171A1 | NM_001010924.2 | NP_001010924.1 | ||
| XM_017015904.2 | XP_016871393.1 | |||
| AL050367.1 | CAB43675.1 | |||
| AY683003.1 | AAV85904.1 | |||
| BC109302.1 | AAI09303.1 | |||
| BC109303.1 | AAI09304.1 | |||
| BX496483.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3622442 | Chr.10:15367490 - 15486645 on Build GRCh38 | Loss |
|
| nsv550017 | Chr.10:15351310 - 15415762 on Build GRCh38 | Loss |
|
| nsv466755 | Chr.10:15334858 - 15477923 on Build GRCh38 | Gain |
|
| nsv5489049 | Chr.10:15367145 - 15487291 on Build GRCh38 | Deletion |
|
| nsv4190122 | Chr.10:15216102 - 15532821 on Build GRCh38 | Duplication |
|
Set Membership: |
|