Search
Search

Entrez Gene ID: | 222389 |
Gene Name: | BEN domain containing 7 |
Gene Aliases: |
C10orf30 |
Location: |
Chr.10:13438481-13529634 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv825252 | Chr.10:13492728 - 13549437 on Build GRCh38 | Gain |
|
| nsv549988 | Chr.10:13511345 - 13563368 on Build GRCh38 | Loss |
|
| nsv6438038 | Chr.10:13520424 - 13522157 on Build GRCh38 | Deletion |
|
| nsv825255 | Chr.10:13509980 - 13543057 on Build GRCh38 | Gain |
|
Set Membership: |
|