Search
Search

Entrez Gene ID: | 222389 |
Gene Name: | BEN domain containing 7 |
Gene Aliases: |
C10orf30 |
Location: |
Chr.10:13438481-13529634 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6450560 | Chr.10:13488699 - 13490974 on Build GRCh38 | Deletion |
|
Set Membership: |
|