Search
Search

Entrez Gene ID: | 8029 |
Gene Name: | cubilin |
Gene Aliases: |
IFCR, MGA1, gp280 |
Location: |
Chr.10:16823966-17130492 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CUBN | NM_001081.3 | NP_001072.2 | ||
| XM_011519708.2 | XP_011518010.1 | |||
| XM_011519709.2 | 1 | 382 | XP_011518011.1 | |
| XM_011519710.2 | XP_011518012.1 | |||
| AF034611.1 | AAC82612.1 | |||
| AK075228.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1039697 | Chr.10:17006669 - 17549229 on Build GRCh38 | Loss |
|
Set Membership: |
|