Search
Search

Entrez Gene ID: | 2894 |
Gene Name: | glutamate ionotropic receptor delta type subunit 1 |
Gene Aliases: |
GluD1, GluD1-b |
Location: |
Chr.10:85599552-86366795 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRID1 | NM_017551.3 | NP_060021.1 | ||
| XM_047425122.1 | XP_047281078.1 | |||
| XM_047425123.1 | XP_047281079.1 | |||
| AB033046.1 | BAA86534.1 | |||
| AK127168.1 | BAG54447.1 | |||
| AK293104.1 | BAF85793.1 | |||
| AK302192.1 | BAH13646.1 | |||
| BC039263.1 | AAH39263.1 | |||
| BK014583.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1052427 | Chr.10:85234813 - 86270793 on Build GRCh38 | Gain |
|
Set Membership: |
|