Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05221468_cn
          See other OGDHL CNV Assays ›
          Gene Symbol
          OGDHL
          Assay Reference Genome
          Location

          Chr.10:49749343 on build GRCh38
          Cytoband
          Assay ID Hs05221468_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          55753

          Gene Name:

          oxoglutarate dehydrogenase L

          Gene Aliases:

          YOBELN

          Location:

          Chr.10:49734641-49762379 on Build GRCh38

          Assay Gene Location:

          Within Intron 10
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          OGDHL NM_001143996.2 NP_001137468.1
          NM_001143997.2 NP_001137469.1
          NM_001347819.1 NP_001334748.1
          NM_001347820.1 NP_001334749.1
          NM_001347821.2 NP_001334750.1
          NM_001347822.1 NP_001334751.1
          NM_001347823.1 NP_001334752.1
          NM_001347824.2 NP_001334753.1
          NM_001347825.2 NP_001334754.1
          NM_001347826.1 NP_001334755.1
          NM_018245.3 NP_060715.2
          NR_144682.2
          NR_144683.2
          NR_144684.2
          NR_144685.2
          NR_144686.2
          XM_011539946.3 XP_011538248.1
          XM_047425466.1 XP_047281422.1
          AB033116.3 BAA86604.3
          AK001713.1 BAA91855.1
          AK225558.1
          AK290226.1 BAF82915.1
          AK296551.1 BAG59174.1
          AK303729.1 BAG64705.1
          AK307759.1
          BC026320.1 AAH26320.1
          DA411851.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv1048905 Chr.10:49720681 - 49811144 on Build GRCh38 Gain OGDHL
          nsv4184636 Chr.10:49712885 - 49815631 on Build GRCh38 Duplication OGDHL
          nsv3241781 Chr.10:49729756 - 49754249 on Build GRCh38 Insertion OGDHL
          nsv1042677 Chr.10:47993784 - 49928329 on Build GRCh38 Loss PGBD3 PARG TMEM273 AGAP12P FRMPD2 ARHGAP22 DRGX C10orf71-AS1 LRRC18 FAM170B-AS1 C10orf71 ERCC6 FAM25C C10orf53 WDFY4 MIR4294 CHAT FAM170B MAPK8 VSTM4 OGDHL SLC18A3 ARHGAP22-IT1

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          glycolytic process
          tricarboxylic acid cycle
          2-oxoglutarate metabolic process
          oxoglutarate dehydrogenase (succinyl-transferring) activity
          protein binding
          oxidoreductase activity
          oxidoreductase activity, acting on the aldehyde or oxo group of donors, disulfide as acceptor
          thiamine pyrophosphate binding
          metal ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline