Search
Search

Entrez Gene ID: | 119180 |
Gene Name: | lysozyme like 2 |
Gene Aliases: |
LYZD2 |
Location: |
Chr.10:30606221-30629753 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LYZL2 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831822 | Chr.10:30459326 - 30628314 on Build GRCh38 | Gain |
|
| nsv1161936 | Chr.10:30113251 - 31078102 on Build GRCh38 | Duplication |
|
| nsv4179249 | Chr.10:30578922 - 30613138 on Build GRCh38 | Deletion |
|
| nsv6445798 | Chr.10:30596728 - 30611959 on Build GRCh38 | Deletion |
|
| nsv6440335 | Chr.10:30579001 - 30613100 on Build GRCh38 | Deletion |
|
| nsv4188826 | Chr.10:30596733 - 30611959 on Build GRCh38 | Deletion |
|