Search
Search

Entrez Gene ID: | 4745 |
Gene Name: | neural EGFL like 1 |
Gene Aliases: |
IDH3GL, NRP1 |
Location: |
Chr.11:20669551-21575686 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NELL1 | NM_001288713.1 | NP_001275642.1 | ||
| NM_001288714.1 | NP_001275643.1 | |||
| NM_006157.4 | NP_006148.2 | |||
| NM_201551.2 | NP_963845.1 | |||
| AK127805.1 | ||||
| AK299558.1 | ||||
| AK303736.1 | ||||
| AK313445.1 | ||||
| BC069674.1 | AAH69674.1 | |||
| BC096100.1 | AAH96100.1 | |||
| BC096101.1 | AAH96101.1 | |||
| BC096102.3 | AAH96102.1 | |||
| BC096103.3 | AAH96103.1 | |||
| D83017.1 | BAA11680.1 | |||
| JX565026.1 | ||||
| U57523.1 | AAB06946.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832084 | Chr.11:20978054 - 21170572 on Build GRCh38 | Gain |
|
| esv3891942 | Chr.11:20978012 - 21000461 on Build GRCh38 | Loss |
|
| nsv1049813 | Chr.11:20929102 - 21260083 on Build GRCh38 | Loss |
|
| esv3625562 | Chr.11:20882377 - 21018715 on Build GRCh38 | Gain |
|
Set Membership: |
|