Search
Search

Entrez Gene ID: | 338645 |
Gene Name: | leucine zipper protein 2 |
Gene Aliases: |
KFSP2566, PRO6246 |
Location: |
Chr.11:24494304-25082640 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LUZP2 | NM_001009909.3 | NP_001009909.2 | ||
| NM_001252008.1 | NP_001238937.1 | |||
| NM_001252010.1 | NP_001238939.1 | |||
| XM_017017648.1 | XP_016873137.1 | |||
| AK126974.1 | ||||
| AK309077.1 | ||||
| AL832641.3 | CAD90000.1 | |||
| BC132829.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2658856 | Chr.11:25008726 - 25038686 on Build GRCh38 | Deletion |
|
| dgv1710n54 | Chr.11:25020209 - 25064963 on Build GRCh38 | Loss |
|
| esv3625685 | Chr.11:25008727 - 25038638 on Build GRCh38 | Loss |
|
| nsv1053055 | Chr.11:24678466 - 25272005 on Build GRCh38 | Gain |
|
| nsv467758 | Chr.11:24947682 - 25039801 on Build GRCh38 | Loss |
|
| dgv2e49 | Chr.11:24781123 - 25069228 on Build GRCh38 | Loss |
|
| nsv832089 | Chr.11:24996525 - 25179086 on Build GRCh38 | Gain |
|
| nsv553827 | Chr.11:24837090 - 25357735 on Build GRCh38 | Gain |
|
| esv2422339 | Chr.11:24983071 - 25525119 on Build GRCh38 | Deletion |
|
| esv3625687 | Chr.11:25030443 - 25038076 on Build GRCh38 | Loss |
|