Search
Search

Entrez Gene ID: | 406965 |
Gene Name: | microRNA 190a |
Gene Aliases: |
MIR190, MIRN190, hsa-mir-190a, miR-190, mir-190a |
Location: |
Chr.15:62823957-62824041 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR190A | NR_029709.1 |
Entrez Gene ID: | 83660 |
Gene Name: | talin 2 |
Gene Aliases: |
ILWEQ |
Location: |
Chr.15:62390527-62844631 on Build GRCh38 |
Assay Gene Location: | Within Intron 62 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TLN2 | NM_015059.2 | NP_055874.2 | ||
| XM_005254708.4 | XP_005254765.1 | |||
| XM_005254710.4 | XP_005254767.1 | |||
| XM_005254711.4 | XP_005254768.1 | |||
| XM_005254712.4 | XP_005254769.1 | |||
| XM_005254713.4 | XP_005254770.1 | |||
| XM_005254714.3 | XP_005254771.1 | |||
| XM_005254715.2 | XP_005254772.1 | |||
| XM_006720717.3 | XP_006720780.1 | |||
| XM_017022665.1 | XP_016878154.1 | |||
| XM_017022666.1 | XP_016878155.1 | |||
| XM_017022667.1 | XP_016878156.1 | |||
| XM_017022668.1 | XP_016878157.1 | |||
| XM_017022669.1 | XP_016878158.1 | |||
| AB002318.2 | BAA20778.2 | |||
| AB621808.1 | ||||
| AF402000.1 | AAM73764.1 | |||
| AK294570.1 | ||||
| BX647654.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1562 | Chr.15:62819554 - 62854951 on Build GRCh38 | Insertion |
|
| nsv1045036 | Chr.15:62725022 - 62891062 on Build GRCh38 | Gain |
|
| nsv833032 | Chr.15:62723242 - 62899303 on Build GRCh38 | Loss |
|
| nsv1035441 | Chr.15:62406037 - 63349997 on Build GRCh38 | Gain |
|
Set Membership: |
|