Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05359942_cn
          See other PAK6 CNV Assays ›
          Gene Symbol
          PAK6
          Assay Reference Genome
          Location

          Chr.15:40265714 on build GRCh38
          Cytoband
          15q15.1
          Assay ID Hs05359942_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          56924

          Gene Name:

          p21 (RAC1) activated kinase 6

          Gene Aliases:

          PAK5

          Location:

          Chr.15:40239091-40277487 on Build GRCh38

          Assay Gene Location:

          Within Intron 4
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PAK6 NM_001276717.1 NP_001263646.1
          NM_001276718.1 NP_001263647.1
          NM_020168.5 NP_064553.1
          AF276893.1 AAF82800.1
          AJ236915.1 CAC18720.1
          AK131522.1
          BC035596.1 AAH35596.1
          DA191028.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv471240 Chr.15:40235954 - 40303426 on Build GRCh38 Loss PLCB2 PAK6 C15orf56 ANKRD63 BUB1B-PAK6
          nsv510660 Chr.15:40260705 - 40429684 on Build GRCh38 Deletion DISP2 PLCB2 PAK6 PHGR1 KNSTRN C15orf52 ANKRD63 INAFM2 BUB1B-PAK6 IVD
          esv34145 Chr.15:40257416 - 40597626 on Build GRCh38 Loss DISP2 PLCB2 PAK6 PHGR1 ANKRD63 RPUSD2 INAFM2 BAHD1 C15orf57 CHST14 KNSTRN C15orf52 KNL1 BUB1B-PAK6 IVD MRPL42P5
          nsv517541 Chr.15:40265067 - 40303426 on Build GRCh38 Loss PLCB2 PAK6 ANKRD63 BUB1B-PAK6
          nsv952611 Chr.15:40237800 - 40284699 on Build GRCh38 Deletion PAK6 C15orf56 ANKRD63 BUB1B-PAK6

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cell cycle
          regulation of transcription, DNA-templated
          apoptotic process
          cytoskeleton organization
          signal transduction by protein phosphorylation
          cell-cell adhesion
          receptor signaling protein serine/threonine kinase activity
          protein binding
          ATP binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline