Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05410446_cn
          See other MGRN1 CNV Assays ›
          Gene Symbol
          MGRN1
          Assay Reference Genome
          Location

          Chr.16:4687077 on build GRCh38
          Cytoband
          16p13.3
          Assay ID Hs05410446_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          23295

          Gene Name:

          mahogunin ring finger 1

          Gene Aliases:

          RNF156

          Location:

          Chr.16:4616493-4690974 on Build GRCh38

          Assay Gene Location:

          Within Exon 19
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          MGRN1 NM_001142289.2 NP_001135761.2
          NM_001142290.2 17 2574 NP_001135762.1
          NM_001142291.2 16 2508 NP_001135763.2
          NM_015246.3 NP_056061.1
          NR_102267.1
          XM_005255217.1 XP_005255274.1
          XM_005255218.1 XP_005255275.1
          XM_005255219.3 XP_005255276.1
          XM_005255220.3 XP_005255277.1
          XM_005255221.3 XP_005255278.1
          XM_011522441.1 XP_011520743.1
          XM_011522442.2 XP_011520744.1
          XM_017023091.1 17 2577 XP_016878580.1
          XM_017023092.1 XP_016878581.1
          XM_017023093.1 XP_016878582.1
          AB011116.1 17 2530
          BC050389.1 AAH50389.1
          EF471397.2

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv1160333 Chr.16:4570296 - 4709737 on Build GRCh38 Deletion NUDT16L1 MIR6769A C16orf96 UBALD1 ANKS3 MGRN1
          nsv1046986 Chr.16:4049000 - 5555721 on Build GRCh38 Gain SMIM22 ROGDI C16orf71 RBFOX1 VASN MIR6769A CORO7-PAM16 PPL UBN1 PAM16 UBALD1 TFAP4 GLYR1 ZNF500 CORO7 MGRN1 NAGPA-AS1 NUDT16L1 LINC01569 ADCY9 SEPT12 C16orf96 GLIS2-AS1 CDIP1 SEC14L5 DNAJA3 NAGPA HMOX2 ANKS3 SRL GLIS2 C16orf89 ALG1 EEF2KMT LOC105371063 NMRAL1
          nsv1050237 Chr.16:4641028 - 4744872 on Build GRCh38 Loss NUDT16L1 MIR6769A C16orf71 ANKS3 MGRN1

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          protein monoubiquitination
          endosome to lysosome transport
          negative regulation of cAMP-mediated signaling
          negative regulation of G-protein coupled receptor protein signaling pathway
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          ligase activity
          ubiquitin protein ligase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline