Search
Search

Entrez Gene ID: | 22980 |
Gene Name: | transcription factor 25 |
Gene Aliases: |
FKSG26, Hulp1, NULP1, PRO2620, hKIAA1049 |
Location: |
Chr.16:89873583-89911384 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv457624 | Chr.16:89738447 - 89915950 on Build GRCh38 | Loss |
|
| esv2762221 | Chr.16:89610380 - 90039318 on Build GRCh38 | Gain+Loss |
|
| nsv573810 | Chr.16:89857498 - 90037251 on Build GRCh38 | Loss |
|
| nsv428331 | Chr.16:89216635 - 90185980 on Build GRCh38 | Gain+Loss |
|
| nsv573799 | Chr.16:89852186 - 90022379 on Build GRCh38 | Gain |
|
| nsv511563 | Chr.16:89904112 - 89909088 on Build GRCh38 | Loss |
|
| nsv509640 | Chr.16:89899749 - 89969797 on Build GRCh38 | Insertion |
|
| nsv482951 | Chr.16:88633593 - 90228345 on Build GRCh38 | Loss |
|
| nsv1066499 | Chr.16:89891957 - 90028891 on Build GRCh38 | Loss |
|
Set Membership: |
|