Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05442221_cn
          See other TCF25 CNV Assays ›
          Gene Symbol
          TCF25
          Assay Reference Genome
          Location

          Chr.16:89904722 on build GRCh38
          Cytoband
          16q24.3
          Assay ID Hs05442221_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          22980

          Gene Name:

          transcription factor 25

          Gene Aliases:

          FKSG26, Hulp1, NULP1, PRO2620, hKIAA1049

          Location:

          Chr.16:89873583-89911384 on Build GRCh38

          Assay Gene Location:

          Within Intron 15
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          TCF25 NM_014972.2 NP_055787.1
          XM_005256297.2 XP_005256354.1
          XM_005256298.2 XP_005256355.1
          XM_005256299.2 XP_005256356.1
          XM_011522956.2 XP_011521258.1
          XM_011522957.2 XP_011521259.1
          XM_011522958.2 XP_011521260.1
          XM_011522959.2 XP_011521261.1
          XM_011522960.2 XP_011521262.1
          XM_011522961.2 XP_011521263.1
          XM_017023047.1 XP_016878536.1
          XM_017023048.1 XP_016878537.1
          XM_017023049.1 XP_016878538.1
          XM_017023050.1 XP_016878539.1
          XM_017023051.1 XP_016878540.1
          XM_017023052.1 XP_016878541.1
          XM_017023053.1 XP_016878542.1
          XM_017023054.1 XP_016878543.1
          XM_017023055.1 XP_016878544.1
          XM_017023056.1 XP_016878545.1
          XM_017023057.1 XP_016878546.1
          XM_017023058.1 XP_016878547.1
          AB028972.1 BAA83001.1
          AF130092.1 AAG35518.1
          AF322111.1 AAG50276.1
          AK095785.1 1 773
          BC000959.1 AAH00959.1
          BC009349.2 AAH09349.1
          BC011884.2 AAH11884.1
          DQ321703.1 ABC55265.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv457624 Chr.16:89738447 - 89915950 on Build GRCh38 Loss SPIRE2 LOC105371419 ZNF276 LOC107984817 TCF25 FANCA
          esv2762221 Chr.16:89610380 - 90039318 on Build GRCh38 Gain+Loss AFG3L1P SPIRE2 CHMP1A CDK10 ZNF276 GAS8-AS1 VPS9D1 SPATA2L CENPBD1 MC1R SPATA33 LOC105371419 DBNDD1 GAS8 VPS9D1-AS1 LOC107984817 TUBB3 TCF25 FANCA DEF8 DPEP1
          nsv573810 Chr.16:89857498 - 90037251 on Build GRCh38 Loss AFG3L1P CENPBD1 MC1R SPIRE2 DBNDD1 GAS8 GAS8-AS1 TUBB3 TCF25 DEF8
          nsv428331 Chr.16:89216635 - 90185980 on Build GRCh38 Gain+Loss TUBB8P7 CHMP1A CDK10 ZNF276 GAS8-AS1 VPS9D1 CPNE7 SPATA2L CENPBD1 MC1R SPATA33 ANKRD11 RPL13 DBNDD1 GAS8 SNORD68 LOC101927817 LOC107984817 TUBB3 LOC105371414 FANCA DEF8 DPEP1 AFG3L1P SPIRE2 URAHP PRDM7 ZNF778 LOC105371419 SPG7 FAM157C VPS9D1-AS1 TCF25 LOC100287036
          nsv573799 Chr.16:89852186 - 90022379 on Build GRCh38 Gain AFG3L1P CENPBD1 MC1R SPIRE2 LOC105371419 DBNDD1 GAS8 TUBB3 TCF25 DEF8
          nsv511563 Chr.16:89904112 - 89909088 on Build GRCh38 Loss TCF25
          nsv509640 Chr.16:89899749 - 89969797 on Build GRCh38 Insertion CENPBD1 MC1R TUBB3 TCF25 DEF8
          nsv482951 Chr.16:88633593 - 90228345 on Build GRCh38 Loss LOC107987238 CHMP1A RNF166 GAS8-AS1 SNAI3 CPNE7 TRAPPC2L LOC105371409 PABPN1L GALNS CENPBD1 MC1R RPL13 APRT DBNDD1 SNORD68 LOC107984817 CDH15 MIR4722 ACSF3 DPEP1 AFG3L1P LOC100129697 LOC101927793 PRDM7 SPG7 TCF25 LOC100289580 CYBA TUBB8P7 LINC00304 CDK10 ZNF276 VPS9D1 SLC22A31 SPATA2L CDT1 SPATA33 ANKRD11 CTU2 GAS8 LOC101927817 TUBB3 LOC105371414 CBFA2T3 FANCA DEF8 SPIRE2 URAHP IL17C LOC400558 MVD SNAI3-AS1 ZNF778 LOC339059 PIEZO1 LOC105371419 FAM157C VPS9D1-AS1 LOC100287036
          nsv1066499 Chr.16:89891957 - 90028891 on Build GRCh38 Loss AFG3L1P CENPBD1 MC1R DBNDD1 GAS8 TUBB3 TCF25 DEF8

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          heart development
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline