Search
Search

Entrez Gene ID: | 55783 |
Gene Name: | cap methyltransferase 2 |
Gene Aliases: |
AFT, FTSJD1, HMTr2, MTr2 |
Location: |
Chr.16:71282300-71290178 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CMTR2 | XM_017023444.1 | 1 | 400 | XP_016878933.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509631 | Chr.16:71279342 - 71336612 on Build GRCh38 | Insertion |
|
| nsv1861 | Chr.16:71279772 - 71298716 on Build GRCh38 | Insertion |
|
Set Membership: |
|