Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05476389_cn
          See other PIK3R6 CNV Assays ›
          Gene Symbol
          PIK3R6
          Assay Reference Genome
          Location

          Chr.17:8805509 on build GRCh38
          Cytoband
          Assay ID Hs05476389_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          146850

          Gene Name:

          phosphoinositide-3-kinase regulatory subunit 6

          Gene Aliases:

          C17orf38, HsT41028, p84 PIKAP, p87(PIKAP), p87PIKAP

          Location:

          Chr.17:8802722-8867677 on Build GRCh38

          Assay Gene Location:

          Within Intron 19
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PIK3R6 NM_001010855.4 NP_001010855.1
          NM_001290211.1 NP_001277140.1
          NR_110865.1
          XM_011523677.3 XP_011521979.1
          XM_011523678.3 XP_011521980.1
          XM_011523680.3 XP_011521982.1
          XM_011523683.3 XP_011521985.1
          XM_011523686.3 XP_011521988.1
          XM_047435443.1 XP_047291399.1
          XM_047435444.1 XP_047291400.1
          XM_047435446.1 XP_047291402.1
          XM_047435447.1 XP_047291403.1
          XM_047435448.1 XP_047291404.1
          XM_047435449.1 XP_047291405.1
          XM_047435450.1 XP_047291406.1
          XM_047435451.1 XP_047291407.1
          XM_047435452.1 XP_047291408.1
          XM_047435453.1 XP_047291409.1
          XM_047435454.1 XP_047291410.1
          XM_047435456.1 XP_047291412.1
          XM_047435457.1 XP_047291413.1
          AK091819.1 BAG52421.1
          AY753192.1 AAV30085.1
          BC125169.1 AAI25170.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv518675 Chr.17:8696386 - 8818290 on Build GRCh38 Gain MFSD6L SPDYE4 PIK3R6 CCDC42
          nsv3218716 Chr.17:8805026 - 8818034 on Build GRCh38 Deletion PIK3R6
          nsv953828 Chr.17:8796283 - 8806582 on Build GRCh38 Deletion MFSD6L PIK3R6
          nsv1969 Chr.17:8780681 - 8825867 on Build GRCh38 Deletion MFSD6L PIK3R6

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          immune response
          G-protein coupled receptor signaling pathway
          regulation of natural killer cell mediated cytotoxicity
          positive regulation of MAP kinase activity
          positive regulation of T cell differentiation
          positive regulation of angiogenesis
          protein binding
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity
          1-phosphatidylinositol-3-kinase regulator activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline