Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05516146_cn
          See other CDK12 CNV Assays ›
          Gene Symbol
          CDK12
          Assay Reference Genome
          Location

          Chr.17:39543691 on build GRCh38
          Cytoband
          Assay ID Hs05516146_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          51755

          Gene Name:

          cyclin dependent kinase 12

          Gene Aliases:

          CRK7, CRKR, CRKRS

          Location:

          Chr.17:39461486-39567559 on Build GRCh38

          Assay Gene Location:

          Within Intron 14
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          CDK12 XM_011524893.3 XP_011523195.1
          XM_011524894.3 XP_011523196.1
          XM_011524895.3 XP_011523197.1
          XM_011524897.3 XP_011523199.1
          XM_011524898.3 XP_011523200.1
          XM_011524906.3 XP_011523208.1
          XM_011524907.3 XP_011523209.1
          XM_017024753.3 XP_016880242.1
          XM_047436255.1 XP_047292211.1
          XM_047436256.1 XP_047292212.1
          XM_047436257.1 XP_047292213.1
          XM_047436258.1 XP_047292214.1
          XM_047436259.1 XP_047292215.1
          XM_047436260.1 XP_047292216.1
          XM_047436261.1 XP_047292217.1
          XM_047436262.1 XP_047292218.1
          XM_047436263.1 XP_047292219.1
          XM_047436264.1 XP_047292220.1
          XM_047436265.1 XP_047292221.1
          XM_047436266.1 XP_047292222.1
          XM_047436267.1 XP_047292223.1
          XM_047436268.1 XP_047292224.1
          XM_047436269.1 XP_047292225.1
          XM_047436270.1 XP_047292226.1
          XM_047436272.1 XP_047292228.1
          XM_047436273.1 XP_047292229.1
          XM_047436274.1 XP_047292230.1
          XM_047436275.1 XP_047292231.1
          XM_047436276.1 XP_047292232.1
          XM_047436277.1 XP_047292233.1
          XM_047436278.1 XP_047292234.1
          XM_047436279.1 XP_047292235.1
          XM_047436280.1 XP_047292236.1
          XM_047436281.1 XP_047292237.1
          XM_047436282.1 XP_047292238.1
          XM_047436283.1 XP_047292239.1
          XM_047436284.1 XP_047292240.1
          XM_047436285.1 XP_047292241.1
          XM_047436286.1 XP_047292242.1
          XM_047436287.1 XP_047292243.1
          XM_047436288.1 XP_047292244.1
          XM_047436289.1 XP_047292245.1
          XM_047436290.1 XP_047292246.1
          XM_047436291.1 XP_047292247.1
          XM_047436292.1 XP_047292248.1
          XM_047436293.1 XP_047292249.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv4254626 Chr.17:39542438 - 39546090 on Build GRCh38 Duplication CDK12
          nsv521294 Chr.17:39538456 - 39648605 on Build GRCh38 Loss PPP1R1B NEUROD2 CDK12 STARD3
          nsv1063848 Chr.17:39419369 - 39551339 on Build GRCh38 Gain MED1 CDK12
          dgv40n68 Chr.17:39536627 - 39726611 on Build GRCh38 Loss PPP1R1B NEUROD2 PNMT TCAP PGAP3 CDK12 STARD3 MIR4728 ERBB2

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription from RNA polymerase II promoter
          mRNA processing
          RNA splicing
          positive regulation of transcription elongation from RNA polymerase II promoter
          negative regulation of intracellular estrogen receptor signaling pathway
          regulation of MAP kinase activity
          negative regulation of MAPK cascade
          positive regulation of transcription from RNA polymerase II promoter
          regulation of cell cycle
          cellular response to estrogen stimulus
          negative regulation of stem cell differentiation
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          cyclin-dependent protein serine/threonine kinase activity
          protein binding
          ATP binding
          RNA polymerase II carboxy-terminal domain kinase activity
          kinase activity
          transferase activity
          protein kinase binding
          cyclin binding
          protein serine kinase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline