Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05560291_cn
          See other TRMU CNV Assays ›
          Gene Symbol
          TRMU
          Assay Reference Genome
          Location

          Chr.22:46337157 on build GRCh38
          Cytoband
          Assay ID Hs05560291_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          55687

          Gene Name:

          tRNA mitochondrial 2-thiouridylase

          Gene Aliases:

          LCAL3, MTO2, MTU1, TRMT, TRMT1

          Location:

          Chr.22:46335714-46357340 on Build GRCh38

          Assay Gene Location:

          Within Intron 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          TRMU NM_001282782.2 NP_001269711.1
          NM_001282783.2 NP_001269712.1
          NM_001282784.2 NP_001269713.1
          NM_001282785.2 NP_001269714.1
          NM_018006.5 NP_060476.2
          NR_104240.2
          NR_104241.2
          XM_011530272.3 XP_011528574.1
          XM_011530273.3 XP_011528575.1
          AB178028.1 BAD66875.1
          AK001002.1 BAA91462.1
          AK290712.1 BAF83401.1
          AK295173.1 BAG58183.1
          AK303020.1 BAG64147.1
          AY062123.2 AAL38183.1
          BC027991.1 AAH27991.1
          BX952612.1
          CN365141.1
          CR456445.1 CAG30331.1
          DQ309998.1 ABC46690.1
          DQ309999.1 ABC46691.1
          DQ310000.1 ABC46692.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv829293 Chr.22:46241054 - 46741639 on Build GRCh38 Gain GTSE1 GRAMD4 CERK LINC02925 CDPF1 CELSR1 PKDREJ PPARA TRMU TTC38 GTSE1-DT
          nsv5541587 Chr.22:46323745 - 46345185 on Build GRCh38 Deletion GTSE1 TRMU
          nsv829281 Chr.22:45861485 - 46735858 on Build GRCh38 Gain GTSE1 LINC02939 PRR34 MIR3619 MIRLET7BHG PPARA WNT7B LOC124905135 TRMU GTSE1-DT GRAMD4 LOC730668 LINC02925 CELSR1 PKDREJ LOC105373071 MIRLET7A3 MIR4763 CERK LOC107985535 CDPF1 MIRLET7B LINC00899 PRR34-AS1 TTC38
          nsv834225 Chr.22:46323584 - 46510799 on Build GRCh38 Loss GTSE1 CELSR1 TRMU
          nsv6555083 Chr.22:46266177 - 46420841 on Build GRCh38 Duplication GTSE1 CELSR1 TRMU TTC38 GTSE1-DT
          nsv471214 Chr.22:46278315 - 46367774 on Build GRCh38 Loss GTSE1 CELSR1 TRMU TTC38 GTSE1-DT
          nsv1066139 Chr.22:46310352 - 46352592 on Build GRCh38 Gain GTSE1 TRMU
          esv3647951 Chr.22:46324981 - 46344914 on Build GRCh38 Loss GTSE1 TRMU

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA wobble position uridine thiolation
          tRNA processing
          tRNA binding
          nucleotide binding
          RNA binding
          protein binding
          ATP binding
          transferase activity
          sulfurtransferase activity
          tRNA-5-taurinomethyluridine 2-sulfurtransferase

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline