Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05562499_cn
          See other LARGE1 CNV Assays ›
          Gene Symbol
          LARGE1
          Assay Reference Genome
          Location

          Chr.22:33820802 on build GRCh38
          Cytoband
          Assay ID Hs05562499_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          9215

          Gene Name:

          LARGE xylosyl- and glucuronyltransferase 1

          Gene Aliases:

          LARGE, MDC1D, MDDGA6, MDDGB6

          Location:

          Chr.22:33066663-33922824 on Build GRCh38

          Assay Gene Location:

          Within Intron 7
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          LARGE1 NM_001362949.2 NP_001349878.1
          NM_001362951.2 NP_001349880.1
          NM_001362953.2 NP_001349882.1
          NM_001378624.1 NP_001365553.1
          NM_001378625.1 NP_001365554.1
          NM_001378626.1 NP_001365555.1
          NM_001378627.1 NP_001365556.1
          NM_001378628.1 NP_001365557.1
          NM_001378629.1 NP_001365558.1
          NM_004737.7 NP_004728.1
          NM_133642.5 NP_598397.1
          XM_024452302.2 XP_024308070.1
          XM_024452303.2 XP_024308071.1
          XM_047441599.1 XP_047297555.1
          XM_047441600.1 XP_047297556.1
          XM_047441601.1 XP_047297557.1
          XM_047441602.1 XP_047297558.1
          XM_047441604.1 XP_047297560.1
          XM_047441605.1 XP_047297561.1
          AB011181.3 BAA25535.3
          AI199197.1
          AJ007583.1 CAA07571.1
          AK294766.1 BAH11875.1
          AK295636.1 BAH12135.1
          BF057493.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv6551065 Chr.22:33808390 - 33826629 on Build GRCh38 Deletion LARGE1
          nsv526550 Chr.22:33808943 - 33825972 on Build GRCh38 Loss LARGE1
          nsv1056001 Chr.22:33810157 - 33825972 on Build GRCh38 Loss LARGE1
          dgv1546n166 Chr.22:33803461 - 33826629 on Build GRCh38 Deletion LARGE1
          nsv459878 Chr.22:33710759 - 33830604 on Build GRCh38 Loss LARGE-AS1 LARGE1
          nsv6548162 Chr.22:33787559 - 33825116 on Build GRCh38 Deletion LARGE1
          nsv6042341 Chr.22:33735842 - 33858438 on Build GRCh38 Deletion LARGE-AS1 LARGE1
          esv2763261 Chr.22:33435362 - 33892401 on Build GRCh38 Loss LARGE-AS1 LARGE1 MIR4764 SNORA50B
          nsv524156 Chr.22:33749609 - 33864750 on Build GRCh38 Loss LARGE-AS1 LARGE1

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel development
          behavioral fear response
          neuron migration
          N-acetylglucosamine metabolic process
          protein glycosylation
          protein O-linked glycosylation
          protein targeting to membrane
          glycosphingolipid biosynthetic process
          water transport
          intracellular protein transport
          striated muscle contraction
          plasma membrane organization
          cytoskeleton organization
          chemical synaptic transmission
          neuromuscular synaptic transmission
          brain development
          heart development
          muscle organ development
          neuromuscular junction development
          sensory perception of sound
          memory
          protein localization
          determination of adult lifespan
          glycoprotein metabolic process
          glycoprotein biosynthetic process
          response to light stimulus
          response to mechanical stimulus
          gene expression
          retina layer formation
          myotube differentiation
          dentate gyrus development
          nerve development
          principal sensory nucleus of trigeminal nerve development
          macrophage differentiation
          post-embryonic hindlimb morphogenesis
          multicellular organism growth
          protein O-linked mannosylation
          protein modification process
          myelination
          skeletal muscle tissue regeneration
          post-translational protein modification
          muscle cell cellular homeostasis
          regulation of synaptic plasticity
          astrocyte differentiation
          cardiac muscle tissue development
          skeletal muscle fiber development
          synapse organization
          neuromuscular process controlling posture
          neuromuscular process
          synaptic growth at neuromuscular junction
          localization of cell
          striated muscle cell development
          cardiac muscle cell development
          transmembrane transport
          retina development in camera-type eye
          limb development
          long-term synaptic potentiation
          bone development
          skeletal muscle organ development
          retina vasculature development in camera-type eye
          connective tissue development
          protein-containing complex assembly
          basement membrane organization
          potassium ion transmembrane transport
          protein localization to membrane
          protein localization to plasma membrane
          walking behavior
          acetylcholine receptor signaling pathway
          skeletal muscle fiber differentiation
          reactive gliosis
          catalytic activity
          protein binding
          acetylglucosaminyltransferase activity
          glucuronosyltransferase activity
          transferase activity
          transferase activity, transferring glycosyl groups
          transferase activity, transferring hexosyl groups
          manganese ion binding
          UDP-xylosyltransferase activity
          xylosyltransferase activity
          metal ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline