Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05571976_cn
          See other TRMU CNV Assays ›
          Gene Symbol
          TRMU
          Assay Reference Genome
          Location

          Chr.22:46346378 on build GRCh38
          Cytoband
          Assay ID Hs05571976_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          55687

          Gene Name:

          tRNA mitochondrial 2-thiouridylase

          Gene Aliases:

          LCAL3, MTO2, MTU1, TRMT, TRMT1

          Location:

          Chr.22:46335714-46357340 on Build GRCh38

          Assay Gene Location:

          Overlaps Intron 3 - Exon 4
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          TRMU NM_001282782.2 NP_001269711.1
          NM_001282783.2 NP_001269712.1
          NM_001282784.2 NP_001269713.1
          NM_001282785.2 NP_001269714.1
          NM_018006.5 NP_060476.2
          NR_104240.2
          NR_104241.2
          XM_011530272.3 XP_011528574.1
          XM_011530273.3 XP_011528575.1
          XM_047441444.1 XP_047297400.1
          XM_047441445.1 XP_047297401.1
          XM_047441446.1 XP_047297402.1
          AB178028.1 BAD66875.1
          AK001002.1 BAA91462.1
          AK290712.1 BAF83401.1
          AK295173.1 BAG58183.1
          AK303020.1 BAG64147.1
          AY062123.2 AAL38183.1
          BC027991.1 AAH27991.1
          BX952612.1
          CN365141.1
          CR456445.1 CAG30331.1
          DQ309998.1 ABC46690.1
          DQ309999.1 ABC46691.1
          DQ310000.1 ABC46692.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv829293 Chr.22:46241054 - 46741639 on Build GRCh38 Gain GTSE1-DT PPARA GTSE1 CERK TRMU TTC38 CDPF1 GRAMD4 PKDREJ LINC02925 CELSR1
          nsv829281 Chr.22:45861485 - 46735858 on Build GRCh38 Gain GTSE1-DT LOC107985535 GTSE1 PKDREJ LOC124905135 TTC38 PRR34 LOC730668 MIR4763 LINC02939 PPARA LOC105373071 LINC00899 TRMU WNT7B MIRLET7B MIRLET7BHG CERK CDPF1 MIR3619 GRAMD4 LINC02925 CELSR1 PRR34-AS1 MIRLET7A3
          nsv834225 Chr.22:46323584 - 46510799 on Build GRCh38 Loss GTSE1 TRMU CELSR1
          nsv6555083 Chr.22:46266177 - 46420841 on Build GRCh38 Duplication GTSE1-DT GTSE1 TRMU TTC38 CELSR1
          nsv471214 Chr.22:46278315 - 46367774 on Build GRCh38 Loss GTSE1-DT GTSE1 TRMU TTC38 CELSR1
          nsv1066139 Chr.22:46310352 - 46352592 on Build GRCh38 Gain GTSE1 TRMU

          Back To Top

          More Information


          Set Membership:

          Intragenic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA wobble position uridine thiolation
          tRNA processing
          tRNA binding
          nucleotide binding
          RNA binding
          protein binding
          ATP binding
          transferase activity
          sulfurtransferase activity
          tRNA-5-taurinomethyluridine 2-sulfurtransferase

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline