Search
Search

Entrez Gene ID: | 1756 |
Gene Name: | dystrophin |
Gene Aliases: |
BMD, CMD3B, DXS142, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, MRX85 |
Location: |
Chr.X:31119219-33339609 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv508n21 | Chr.X:33047771 - 33302495 on Build GRCh38 | Gain |
|
| nsv522546 | Chr.X:33057861 - 33095530 on Build GRCh38 | Gain |
|
Set Membership: |
|