Search
Search

Entrez Gene ID: | 51284 |
Gene Name: | toll like receptor 7 |
Gene Aliases: |
TLR7-like |
Location: |
Chr.X:12867083-12890361 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TLR7 | NM_016562.3 | NP_057646.1 | ||
| AF240467.1 | AAF60188.1 | |||
| AF245702.1 | AAF78035.1 | |||
| AK313858.1 | ||||
| AY358292.1 | AAQ88659.1 | |||
| BC033651.1 | AAH33651.1 |
Set Membership: |
|