Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05804093_cn
          See other MARK1 CNV Assays ›
          Gene Symbol
          MARK1
          Assay Reference Genome
          Location

          Chr.1:220601918 on build GRCh38
          Cytoband
          Assay ID Hs05804093_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          4139

          Gene Name:

          microtubule affinity regulating kinase 1

          Gene Aliases:

          MARK, Par-1c, Par1c

          Location:

          Chr.1:220528136-220664461 on Build GRCh38

          Assay Gene Location:

          Within Intron 8
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          MARK1 NM_001286124.2 NP_001273053.1
          NM_001286126.2 NP_001273055.1
          NM_001286128.2 NP_001273057.1
          NM_018650.5 NP_061120.3
          XM_005273134.6 XP_005273191.1
          XM_006711326.5 XP_006711389.1
          XM_011509561.4 XP_011507863.1
          XM_011509562.4 XP_011507864.1
          XM_017001305.3 XP_016856794.1
          XM_024447090.2 XP_024302858.1
          XM_047420844.1 XP_047276800.1
          AB040910.1 BAA96001.2
          AF154845.1 AAF72103.1
          AK027493.1 BAB55152.1
          AK295506.1 BAG58425.1
          AK314614.1
          BC113869.1 AAI13870.1
          BC114478.1 AAI14479.1
          KJ534879.1 AHW56519.1
          KJ534880.1 AHW56520.1
          KJ534881.1 AHW56521.1
          KJ535089.1 AHW56728.1

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic non-DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          neuron migration
          protein phosphorylation
          cytoskeleton organization
          positive regulation of gene expression
          negative regulation of gene expression
          negative regulation of epithelial to mesenchymal transition
          regulation of neuron projection development
          Wnt signaling pathway
          intracellular signal transduction
          regulation of dendrite development
          establishment of mitochondrion localization
          regulation of postsynapse assembly
          nucleotide binding
          magnesium ion binding
          phosphatidylserine binding
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          phosphatidylinositol-4,5-bisphosphate binding
          lipid binding
          kinase activity
          transferase activity
          metal ion binding
          tau protein binding
          tau-protein kinase activity
          phosphatidic acid binding
          protein serine kinase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline