Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05913021_cn
          See other CAMK2D CNV Assays ›
          Gene Symbol
          CAMK2D
          Assay Reference Genome
          Location

          Chr.4:113749390 on build GRCh38
          Cytoband
          4q26
          Assay ID Hs05913021_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          817

          Gene Name:

          calcium/calmodulin dependent protein kinase II delta

          Gene Aliases:

          CAMKD

          Location:

          Chr.4:113451032-113761927 on Build GRCh38

          Assay Gene Location:

          Within Intron 3
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          CAMK2D NM_001221.3 NP_001212.2
          NM_001321566.1 NP_001308495.1
          NM_001321567.1 NP_001308496.1
          NM_001321568.1 NP_001308497.1
          NM_001321569.1 NP_001308498.1
          NM_001321570.1 NP_001308499.1
          NM_001321571.1 NP_001308500.1
          NM_001321572.1 NP_001308501.1
          NM_001321573.1 NP_001308502.1
          NM_001321574.1 NP_001308503.1
          NM_001321575.1 NP_001308504.1
          NM_001321576.1 NP_001308505.1
          NM_001321577.1 NP_001308506.1
          NM_001321578.1 NP_001308507.1
          NM_001321579.1 NP_001308508.1
          NM_001321580.1 NP_001308509.1
          NM_001321581.1 NP_001308510.1
          NM_001321582.1 NP_001308511.1
          NM_001321583.1 NP_001308512.1
          NM_001321584.1 NP_001308513.1
          NM_001321585.1 NP_001308514.1
          NM_001321586.1 NP_001308515.1
          NM_001321587.1 NP_001308516.1
          NM_001321588.1 NP_001308517.1
          NM_001321589.1 NP_001308518.1
          NM_001321590.1 NP_001308519.1
          NM_001321591.1 NP_001308520.1
          NM_172114.1 NP_742112.1
          NM_172115.2 NP_742113.1
          NM_172127.2 NP_742125.1
          NM_172128.2 NP_742126.1
          NM_172129.1 NP_742127.1
          XM_005263253.3 XP_005263310.1
          XM_005263254.3 XP_005263311.1
          XM_006714330.2 XP_006714393.1
          XM_011532289.1 XP_011530591.1
          XM_011532290.1 XP_011530592.1
          XM_011532291.1 XP_011530593.1
          XM_011532295.1 XP_011530597.1
          XM_017008672.1 XP_016864161.1
          XM_017008673.1 XP_016864162.1
          XM_017008674.1 XP_016864163.1
          XM_017008675.1 XP_016864164.1
          XM_017008677.1 XP_016864166.1
          XM_017008678.1 XP_016864167.1
          AB209288.1 BAD92525.1
          AF071569.1 AAD20442.1
          AJ252239.1 CAB65123.1
          AK055642.1
          AK294517.1
          AY987011.1 AAX88806.1
          BC022558.1 AAH22558.1
          BC032784.1 AAH32784.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv3601817 Chr.4:113722759 - 113853762 on Build GRCh38 Gain CAMK2D
          nsv525087 Chr.4:113740525 - 114390328 on Build GRCh38 Gain CAMK2D ARSJ

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          MAPK cascade
          regulation of cell growth
          response to hypoxia
          regulation of the force of heart contraction
          regulation of membrane depolarization
          regulation of transcription from RNA polymerase II promoter
          protein phosphorylation
          calcium ion transport
          regulation of heart contraction
          regulation of G2/M transition of mitotic cell cycle
          positive regulation of cardiac muscle hypertrophy
          regulation of cell communication by electrical coupling
          positive regulation of cardiac muscle cell apoptotic process
          regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
          positive regulation of smooth muscle cell migration
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          cellular potassium ion homeostasis
          endoplasmic reticulum calcium ion homeostasis
          positive regulation of Rac protein signal transduction
          positive regulation of GTPase activity
          protein autophosphorylation
          positive regulation of smooth muscle cell proliferation
          protein oligomerization
          relaxation of cardiac muscle
          regulation of ryanodine-sensitive calcium-release channel activity
          interferon-gamma-mediated signaling pathway
          regulation of cellular localization
          cell growth involved in cardiac muscle cell development
          positive regulation of ERK1 and ERK2 cascade
          cellular response to calcium ion
          cardiac muscle cell contraction
          regulation of heart rate by cardiac conduction
          regulation of cardiac muscle cell action potential
          regulation of cardiac muscle cell action potential involved in regulation of contraction
          regulation of cellular response to heat
          regulation of histone deacetylase activity
          regulation of cell communication by electrical coupling involved in cardiac conduction
          regulation of relaxation of cardiac muscle
          negative regulation of sodium ion transmembrane transport
          regulation of calcium ion transmembrane transport via high voltage-gated calcium channel
          negative regulation of sodium ion transmembrane transporter activity
          protein serine/threonine kinase activity
          calmodulin-dependent protein kinase activity
          Ras guanyl-nucleotide exchange factor activity
          protein binding
          calmodulin binding
          ATP binding
          sodium channel inhibitor activity
          titin binding
          protein homodimerization activity
          ion channel binding
          nitric-oxide synthase binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline