Search
Search

Entrez Gene ID: | 4790 |
Gene Name: | nuclear factor kappa B subunit 1 |
Gene Aliases: |
CVID12, EBP-1, KBF1, NF-kB1, NF-kappa-B, NF-kappaB, NFKB-p105, NFKB-p50, NFkappaB, p105, p50 |
Location: |
Chr.4:102501329-102617302 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFKB1 | NM_001165412.1 | NP_001158884.1 | ||
| NM_001319226.1 | NP_001306155.1 | |||
| NM_003998.3 | NP_003989.2 | |||
| XM_011532006.2 | XP_011530308.1 | |||
| XM_011532008.2 | XP_011530310.1 | |||
| AK291450.1 | ||||
| BC051765.1 | AAH51765.1 | |||
| CR749522.1 | CAH18336.1 | |||
| M55643.1 | AAA36361.1 | |||
| M58603.1 | AAA36360.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830020 | Chr.4:102466186 - 102660127 on Build GRCh38 | Loss |
|
Set Membership: |
|