Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05967984_cn
          See other CAMK2D CNV Assays ›
          Gene Symbol
          CAMK2D
          Assay Reference Genome
          Location

          Chr.4:113704107 on build GRCh38
          Cytoband
          Assay ID Hs05967984_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          817

          Gene Name:

          calcium/calmodulin dependent protein kinase II delta

          Gene Aliases:

          CAMKD

          Location:

          Chr.4:113451032-113761738 on Build GRCh38

          Assay Gene Location:

          Within Intron 3
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          CAMK2D NM_001221.4 NP_001212.2
          NM_001321566.2 NP_001308495.1
          NM_001321567.2 NP_001308496.1
          NM_001321568.2 NP_001308497.1
          NM_001321569.2 NP_001308498.1
          NM_001321570.2 NP_001308499.1
          NM_001321571.2 NP_001308500.1
          NM_001321572.2 NP_001308501.1
          NM_001321573.2 NP_001308502.1
          NM_001321574.2 NP_001308503.1
          NM_001321575.2 NP_001308504.1
          NM_001321576.2 NP_001308505.1
          NM_001321577.2 NP_001308506.1
          NM_001321578.2 NP_001308507.1
          NM_001321579.2 NP_001308508.1
          NM_001321580.2 NP_001308509.1
          NM_001321581.2 NP_001308510.1
          NM_001321582.2 NP_001308511.1
          NM_001321583.2 NP_001308512.1
          NM_001321584.2 NP_001308513.1
          NM_001321585.2 NP_001308514.1
          NM_001321586.2 NP_001308515.1
          NM_001321587.2 NP_001308516.1
          NM_001321588.2 NP_001308517.1
          NM_001321589.2 NP_001308518.1
          NM_001321590.2 NP_001308519.1
          NM_001321591.2 NP_001308520.1
          NM_001396964.1 NP_001383893.1
          NM_001396965.1 NP_001383894.1
          NM_001396966.1 NP_001383895.1
          NM_001396967.1 NP_001383896.1
          NM_001399855.1 NP_001386784.1
          NM_001399856.1 NP_001386785.1
          NM_001399857.1 NP_001386786.1
          NM_001399858.1 NP_001386787.1
          NM_001399859.1 NP_001386788.1
          NM_001399860.1 NP_001386789.1
          NM_001399861.1 NP_001386790.1
          NM_001399862.1 NP_001386791.1
          NM_001399863.1 NP_001386792.1
          NM_001399864.1 NP_001386793.1
          NM_001399865.1 NP_001386794.1
          NM_172114.2 NP_742112.1
          NM_172115.3 NP_742113.1
          NM_172127.3 NP_742125.1
          NM_172128.3 NP_742126.1
          NM_172129.2 NP_742127.1
          XM_011532295.3 XP_011530597.1
          XM_017008674.3 XP_016864163.1
          XM_047416220.1 XP_047272176.1
          XM_047416221.1 XP_047272177.1
          XM_047416222.1 XP_047272178.1
          XM_047416223.1 XP_047272179.1
          AB209288.1 BAD92525.1
          AF071569.1 AAD20442.1
          AJ252239.1 CAB65123.1
          AK055642.1 BAG51545.1
          AK294517.1 BAH11796.1
          AY987011.1 AAX88806.1
          BC022558.1 AAH22558.1
          BC032784.1 AAH32784.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv4475 Chr.4:113668443 - 113710725 on Build GRCh38 Deletion CAMK2D
          nsv5547030 Chr.4:68996170 - 113894177 on Build GRCh38 Insertion CFAP299 MIR302B EGF LOC105377267 UTP3 USO1 ADH5 STBD1 TET2-AS1 PPBPP2 BMP3 GSTCD NUDT9 LOC100421494 TSPAN5 DMP1 BMP2K ATOH1 ANKRD17-DT AMTN PRDM8 MMRN1 MIR1255A LOC124900868 ARHGAP24 RPL34-DT MIR575 ENOPH1 AFP PYURF SCARB2 MIR4451 MUC7 CSN2 GIMD1 NKX6-1 ARHGEF38-IT1 ADH7 SNORA101A ADH6 LINC00575 HERC5 LOC105377329 LEF1-AS1 UGT2A1 UGT2B11 INKILN AFF1 TSPAN5-DT CCSER1 WDFY3 PIGY COX18 SMR3A GSTCD-AS1 ANK2 TACR3-AS1 UBE2D3-AS1 NAA11 KLHL8 GRSF1 MRPS18C MIR302C DDIT4L-AS1 SEC24B HTN3 PARM1-AS1 SOWAHB STPG2-AS1 CISD2 PPP3CA-DT COQ2 ARHGEF38 ZGRF1 SMARCAD1 HERC6 RAP1GDS1 MIR302CHG CAMK2D AMBN CXCL6 ALPK1 THAP6 RPS26P25 LOC124900752 CXCL8 PKD2 LAMTOR3 MIR8082 NAP1L5 MIR5705 SNCA CDS1 PTPN13 SEPTIN11 LIN54 GC DKK2 MANBA NFKB1 H2AZ1 AREG BDH2 PPP3CA DMP1-AS1 NUP54 EPGN LOC105377366 CSN1S2BP AIMP1 TBCK MIR4452 PITX2 GRID2 CXXC4-AS1 FAM47E LOC105377352 TIFA MIR576 ANTXR2 PPBP SMARCAD1-DT MTHFD2L CENPE RRH SEC24B-AS1 SPP1 PDHA2 STATH CFI SMR3B AP1AR LOC100507053 HNRNPDL LINC02483 ADH1C LRIT3 THAP9-AS1 MIR297 PCNAP1 HSD17B11 SNCA-AS1 PAPSS1 TET2 TRMT10A AFF1-AS1 MIR302A DCK COL25A1-DT WDFY3-AS1 ADH1B COPS4 MEPE MCUB ODAM PCAT4 GK2 OPRPN NEUROG2-AS1 LINC01218 HTN1 METAP1 LINC01088 UGT2B7 LOC101928893 CXCL2 LOC101929621 SHROOM3 SLC4A4 LOC124900866 INTS12 LOC124900855 ETNPPL PARM1 SNORD144 CDKL2 MIR8066 CXCL1 HERC3 PF4V1 PANCR BANK1 FGF5 SLC9B1 CXCL3 GAR1 UNC5C MIR4450 MIR548AH BMPR1B NPNT PPEF2 PAQR3 FAM47E-STBD1 MTTP ADAMTS3 PIGY-DT LOC102723704 IBSP LNCPRESS2 FAM241A MIR1243 PPM1K UGT2A2 ENAM FAM13A AFM MIR3684 PPA2 ANXA3 FDCSP MAPK10 MRPL1 UGT2B4 HSP90AB3P MIR367 HPGDS CXCL10 PLAC8 SCPPPQ1 RUFY3 LINC01216 SDAD1-AS1 MOB1B UGT2B28 LINC02173 LINC02994 LINC01217 NFKB1-AS1 LINC02499 FRAS1 PRR27 CXCL13 EMCN UBE2D3 SNORD143 ABRAXAS1 UNC5C-AS1 SDAD1 FAM13A-AS1 LINC02945 LEF1 RCHY1 CXXC4 HADH LINC01094 BMP2K-DT C4orf36 EREG LINC00989 HNRNPD-DT CYP2U1-AS1 LINC02428 CNOT6L TMSB4XP8 HELQ CYP2U1 GPRIN3 LARP7 PRDM8-AS1 SLC9B2 ABCG2 CASP6 ANKRD17 SULT1E1 ALB PPM1K-DT RPS6P6 GPAT3 RASGEF1B OSTC LOC124900867 CSN1S1 LINC02267 MAPK10-AS1 CHCHD2P7 EIF4E SGMS2 SCD5 CABS1 CCNG2 MIR302D ENPEP BMPR1B-DT LOC101929577 WDFY3-AS2 COL25A1 LINC02562 CXCL11 TMEM150C CSN1S2AP AP1AR-DT G3BP2 RPL34 PRKG2 STPG2 ELOVL6 JCHAIN ADH4 TIGD2 LINC02503 LOC102725220 CCNI ART3 DDIT4L TACR3 CCDC158 NAAA BTC PLA2G12A SNORD161 CXCL9 NEUROG2 LOC105377276 SLC39A8 PRKG2-AS1 H2AZ1-DT DNAJB14 PDLIM5 C4orf17 SEC31A THAP9 HNRNPD ODAPH CSN3 GAR1-DT UMLILO C4orf54 SULT1B1 CXCL5 RASSF6 SHROOM3-AS1 HSD17B13 HPSE ADH1A DSPP PF4 NPFFR2 SPARCL1 SLC10A6 DAPP1
          nsv5460550 Chr.4:113651381 - 113855381 on Build GRCh38 Duplication CAMK2D

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          regulation of the force of heart contraction
          regulation of membrane depolarization
          regulation of transcription from RNA polymerase II promoter
          protein phosphorylation
          regulation of heart contraction
          positive regulation of cardiac muscle hypertrophy
          regulation of cell communication by electrical coupling
          positive regulation of cardiac muscle cell apoptotic process
          regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
          endoplasmic reticulum calcium ion homeostasis
          regulation of neuronal synaptic plasticity
          relaxation of cardiac muscle
          long-term synaptic potentiation
          regulation of ryanodine-sensitive calcium-release channel activity
          regulation of cellular localization
          cellular response to calcium ion
          cardiac muscle cell contraction
          regulation of heart rate by cardiac conduction
          regulation of cardiac muscle cell action potential
          regulation of cardiac muscle cell action potential involved in regulation of contraction
          regulation of cell communication by electrical coupling involved in cardiac conduction
          regulation of relaxation of cardiac muscle
          negative regulation of sodium ion transmembrane transport
          regulation of calcium ion transmembrane transport via high voltage-gated calcium channel
          regulation of protein localization to plasma membrane
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          calmodulin-dependent protein kinase activity
          protein binding
          calmodulin binding
          ATP binding
          kinase activity
          transferase activity
          sodium channel inhibitor activity
          titin binding
          identical protein binding
          protein homodimerization activity
          ion channel binding
          protein serine kinase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline