Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs06136028_cn
          See other GCNT2 CNV Assays ›
          Gene Symbol
          GCNT2
          Assay Reference Genome
          Location

          Chr.6:10614140 on build GRCh38
          Cytoband
          Assay ID Hs06136028_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          2651

          Gene Name:

          glucosaminyl (N-acetyl) transferase 2 (I blood group)

          Gene Aliases:

          CCAT, CTRCT13, GCNT2C, GCNT5, IGNT, II, NACGT1, NAGCT1, ULG3, bA360O19.2, bA421M1.1

          Location:

          Chr.6:32235-140252 on Build GRCh38

          Assay Gene Location:

          Within Intron 7
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          GCNT2 NM_001374747.1 NP_001361676.1
          NM_001491.3 NP_001482.1
          NM_145649.5 NP_663624.1
          NM_145655.4 NP_663630.2
          XM_005248999.3 XP_005249056.1
          XM_006715052.4 XP_006715115.1
          XM_047418633.1 XP_047274589.1
          AB078432.1 BAC66781.1
          AB078433.1 BAC66782.1
          AF401652.1 AAL50561.1
          AF401653.1 AAL50562.1
          AF458024.1 AAM73864.1
          AF458025.1 AAM73865.1
          AF458026.1 AAM73866.1
          AK090483.1 BAC03464.1
          AK098105.1 BAC05234.1
          AK291195.1 BAF83884.1
          AK291767.1 BAF84456.1
          AK307329.1
          AK307337.1
          AK307373.1
          AK313426.1 BAG36218.1
          AK313903.1 BAG36626.1
          AL832714.1
          AL832719.1
          AY435145.1 AAR95646.1
          AY435146.1 AAR95647.1
          AY435147.1 AAR95648.1
          BC074801.2 AAH74801.1
          BC074802.2 AAH74802.1
          BC130524.1 AAI30525.1
          BX647576.1 CAI46081.1
          BX647668.1
          L19659.1 AAA81777.1
          Z19550.1 CAA79610.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv3113907 Chr.6:10481673 - 10698466 on Build GRCh38 Deletion GCNT2 C6orf52 LINC02522 PAK1IP1
          nsv6399788 Chr.6:10495714 - 10615476 on Build GRCh38 Deletion GCNT2
          nsv600988 Chr.6:10501021 - 10614845 on Build GRCh38 Loss GCNT2
          nsv966670 Chr.6:10447438 - 10871656 on Build GRCh38 Duplication GCNT2 MAK TMEM14B-DT TMEM14B C6orf52 LINC02522 MIR5689HG PAK1IP1 TMEM14C
          nsv600979 Chr.6:10449025 - 10839526 on Build GRCh38 Gain GCNT2 MAK TMEM14B-DT TMEM14B C6orf52 LINC02522 MIR5689HG PAK1IP1 TMEM14C
          nsv6404362 Chr.6:10545723 - 10821818 on Build GRCh38 Duplication GCNT2 MAK TMEM14B-DT TMEM14B C6orf52 PAK1IP1 TMEM14C
          nsv470800 Chr.6:10538711 - 10863092 on Build GRCh38 Gain GCNT2 MAK TMEM14B-DT TMEM14B C6orf52 PAK1IP1 TMEM14C
          nsv6403222 Chr.6:10465448 - 10718419 on Build GRCh38 Deletion GCNT2 C6orf52 LINC02522 PAK1IP1
          esv2759406 Chr.6:10443564 - 10642899 on Build GRCh38 Loss GCNT2 LINC02522 MIR5689HG

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          glycosaminoglycan biosynthetic process
          protein glycosylation
          transforming growth factor beta receptor signaling pathway
          multicellular organism development
          positive regulation of cell proliferation
          glycoprotein biosynthetic process
          posttranscriptional regulation of gene expression
          positive regulation of epithelial to mesenchymal transition
          negative regulation of cell-substrate adhesion
          positive regulation of cell migration
          positive regulation of heterotypic cell-cell adhesion
          maintenance of lens transparency
          positive regulation of protein kinase B signaling
          positive regulation of ERK1 and ERK2 cascade
          protein binding
          N-acetyllactosaminide beta-1,6-N-acetylglucosaminyltransferase activity
          UDP-glycosyltransferase activity
          acetylglucosaminyltransferase activity
          transferase activity
          transferase activity, transferring glycosyl groups

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline