Search
Search

Entrez Gene ID: | 57120 |
Gene Name: | golgi associated PDZ and coiled-coil motif containing |
Gene Aliases: |
CAL, FIG, GOPC1, PIST, dJ94G16.2 |
Location: |
Chr.6:117560269-117602542 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOPC | NM_001017408.2 | NP_001017408.1 | ||
| NM_020399.3 | NP_065132.1 | |||
| AB209385.1 | BAD92622.1 | |||
| AF287894.1 | AAG00572.1 | |||
| AF450008.1 | AAL47160.1 | |||
| AK314486.1 | ||||
| AY033606.1 | AAK57733.1 | |||
| BC009553.2 | AAH09553.1 | |||
| BX419592.2 | ||||
| KF420122.1 | ||||
| KF420123.1 |
Set Membership: |
|