Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs06281480_cn
          See other PTPRJ CNV Assays ›
          Gene Symbol
          PTPRJ
          Assay Reference Genome
          Location

          Chr.11:48102911 on build GRCh38
          Cytoband
          11p11.2
          Assay ID Hs06281480_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          5795

          Gene Name:

          protein tyrosine phosphatase, receptor type J

          Gene Aliases:

          CD148, DEP1, HPTPeta, R-PTP-ETA, SCC1

          Location:

          Chr.11:47980504-48170842 on Build GRCh38

          Assay Gene Location:

          Within Intron 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PTPRJ NM_001098503.1 NP_001091973.1
          NM_002843.3 NP_002834.3
          XM_017018083.1 XP_016873572.1
          XM_017018084.1 XP_016873573.1
          XM_017018085.1 XP_016873574.1
          AL359057.1 CAB94390.1
          BC063417.1 AAH63417.1
          D37781.1 BAA07035.1
          DR007037.1
          U10886.1 AAB36687.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv1111n100 Chr.11:47653918 - 48644472 on Build GRCh38 Gain OR4X1 AGBL2 NUP160 PTPRJ FNBP4 OR4X2 OR4C3 OR4B1 MIR3161 OR4S1 OR4A47
          dgv1112n100 Chr.11:48056312 - 48871655 on Build GRCh38 Gain OR4X1 PTPRJ OR4X2 OR4C3 OR4B1 MIR3161 OR4S1 OR4A47
          nsv1053528 Chr.11:48064599 - 48104179 on Build GRCh38 Loss PTPRJ MIR3161
          esv3891986 Chr.11:48021501 - 49282806 on Build GRCh38 Gain OR4X1 TRIM49B PTPRJ OR4X2 OR4B1 TRIM64C OR4S1 TRIM51CP OR4C3 TRIM53CP MIR3161 FOLH1 OR4A47
          nsv554211 Chr.11:48084934 - 48673319 on Build GRCh38 Gain OR4X1 PTPRJ OR4X2 OR4C3 OR4B1 MIR3161 OR4S1 OR4A47

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          extracellular matrix protein transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of cell proliferation
          negative regulation of platelet-derived growth factor receptor signaling pathway
          regulation of cell adhesion
          negative regulation of cell growth
          negative regulation of cell migration
          peptidyl-tyrosine dephosphorylation
          negative regulation of epidermal growth factor receptor signaling pathway
          negative regulation of vascular permeability
          negative regulation of MAP kinase activity
          positive regulation of cell adhesion
          platelet-derived growth factor receptor signaling pathway
          T cell receptor signaling pathway
          negative regulation of T cell receptor signaling pathway
          positive chemotaxis
          positive regulation of focal adhesion assembly
          positive regulation of protein kinase B signaling
          negative regulation of protein kinase B signaling
          contact inhibition
          protein tyrosine phosphatase activity
          platelet-derived growth factor receptor binding
          protein binding
          beta-catenin binding
          phosphatase activity
          protein kinase binding
          gamma-catenin binding
          mitogen-activated protein kinase binding
          delta-catenin binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline