Search
Search

Entrez Gene ID: | 7334 |
Gene Name: | ubiquitin conjugating enzyme E2 N |
Gene Aliases: |
HEL-S-71, UBC13, UBCHBEN, UBCHBEN; UBC13, UbcH-ben, UbcH13 |
Location: |
Chr.12:93405684-93441947 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2N | NM_003348.4 | NP_003339.1 | ||
| BC000396.2 | AAH00396.1 | |||
| BC003365.1 | AAH03365.1 | |||
| BC108704.1 | AAI08705.1 | |||
| BI459891.1 | ||||
| BT006873.1 | AAP35519.1 | |||
| D83004.1 | BAA11675.1 | |||
| FJ224354.1 | ACI46046.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1654n223 | Chr.12:93217272 - 93436984 on Build GRCh38 | Duplication |
|
Set Membership: |
|