Search
Search

Entrez Gene ID: | 6128 |
Gene Name: | ribosomal protein L6 |
Gene Aliases: |
L6, SHUJUN-2, TAXREB107, TXREB1 |
Location: |
Chr.12:112405181-112418850 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL6 | NM_001320141.1 | 2 | 214 | NP_001307070.1 |
| NM_001320142.1 | 1 | 98 | NP_001307071.1 | |
| XM_017019781.1 | 2 | 1249 | XP_016875270.1 | |
| XM_017019782.1 | 2 | 410 | XP_016875271.1 | |
| BG722198.1 | 2 | 214 | ||
| HY038019.1 | 2 | 214 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3580466 | Chr.12:112403584 - 112419956 on Build GRCh38 | Loss |
|
Set Membership: |
|